Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYC SIRNA Assays ›
Gene Symbol:
Myc
Gene Name:
myelocytomatosis oncogene
Gene Aliases:
RNCMYC, c-myc, mMyc
Chromosome Location:
Chr.7: 102586313 - 102591240 on Build Rnor_6.0
UniGene ID:
Rn.12072
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
47943
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Myc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012603.2
NP_036735.2
2
667
GenBank mRNA
AY679729.1
-
2
504
AY679730.1
-
2
667
BC091699.1
-
2
473
7303974
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001177353, NM_001177354, NM_001177352, NM_010849, AK087961, and others.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
branching involved in ureteric bud morphogenesis
in utero embryonic development
B cell apoptotic process
positive regulation of mesenchymal cell proliferation
regulation of oxidative phosphorylation
positive regulation of B cell apoptotic process
glucose metabolic process
chromatin remodeling
transcription, DNA-templated
DNA-templated transcription, initiation
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
pyruvate transport
amino acid transport
cellular iron ion homeostasis
activation of cysteine-type endopeptidase activity involved in apoptotic process
cellular response to DNA damage stimulus
inner mitochondrial membrane organization
cell cycle arrest
regulation of mitotic cell cycle
cell proliferation
positive regulation of cell proliferation
intrinsic apoptotic signaling pathway in response to DNA damage
glycoprotein metabolic process
response to radiation
pathogenesis
response to wounding
flavonoid metabolic process
response to gamma radiation
regulation of gene expression
negative regulation of gene expression
positive regulation of mitochondrial membrane potential
Wnt signaling pathway
protein processing
transformation of host cell by virus
hypothalamus development
negative regulation of protein binding
regulation of telomere maintenance
response to estradiol
cellular response to UV
cellular response to drug
skeletal muscle cell differentiation
middle ear morphogenesis
response to drug
regulation of apoptotic process
positive regulation of catalytic activity
response to alkaloid
positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
pigmentation
canonical Wnt signaling pathway involved in positive regulation of wound healing
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
G0 to G1 transition
negative regulation of monocyte differentiation
positive regulation of cell cycle
positive regulation of glycolytic process
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of glucose import
lactic acid secretion
positive regulation of fibroblast proliferation
negative regulation of fibroblast proliferation
skeletal system morphogenesis
positive regulation of epithelial cell proliferation
detection of mechanical stimulus involved in sensory perception of sound
chromosome organization
negative regulation of cell division
canonical Wnt signaling pathway
negative regulation of cell death
cellular response to carbohydrate stimulus
cellular response to interleukin-1
cellular response to estrogen stimulus
cellular response to organic cyclic compound
cellular response to hydrostatic pressure
fungal-type cell wall polysaccharide metabolic process
positive regulation of metanephric cap mesenchymal cell proliferation
positive regulation of cellular respiration
positive regulation of DNA biosynthetic process
positive regulation of response to DNA damage stimulus
positive regulation of ATP biosynthetic process
positive regulation of apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
transcription factor binding
protein complex binding
sequence-specific DNA binding
protein heterodimerization activity
protein dimerization activity
repressing transcription factor binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0