Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SPARC SIRNA Assays ›
Gene Symbol:
Sparc
Gene Name:
secreted protein, acidic, cysteine-rich (osteonectin)
Gene Aliases:
-
Chromosome Location:
Chr.10: 40742390 - 40764222 on Build Rnor_6.0
UniGene ID:
Rn.234316
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48083
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Sparc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012656.1
NP_036788.1
5
225
XM_008767663.1
XP_008765885.1
4
303
GenBank mRNA
BC061777.1
-
4
240
5621666
FQ212364.1
-
4
294
FQ212499.1
-
4
259
FQ213815.1
-
4
259
FQ214492.1
-
4
259
FQ214649.1
-
4
259
FQ214673.1
-
4
259
FQ215195.1
-
4
258
FQ215581.1
-
4
260
FQ215622.1
-
4
261
FQ215818.1
-
4
270
FQ215935.1
-
4
259
FQ216823.1
-
4
261
FQ217546.1
-
4
260
FQ219866.1
-
4
241
FQ219932.1
-
5
241
FQ219983.1
-
4
259
FQ220003.1
-
4
259
FQ220043.1
-
4
257
FQ220046.1
-
4
259
FQ220186.1
-
4
257
FQ220279.1
-
2
83
FQ220287.1
-
4
261
FQ220443.1
-
4
241
FQ220547.1
-
4
259
FQ220572.1
-
4
259
FQ220644.1
-
4
259
FQ220827.1
-
4
259
FQ220880.1
-
4
259
FQ220901.1
-
4
259
FQ222224.1
-
4
259
FQ223025.1
-
4
259
FQ225033.1
-
4
261
FQ225092.1
-
4
255
FQ225200.1
-
4
259
FQ225220.1
-
4
238
FQ225486.1
-
4
241
FQ225609.1
-
4
259
FQ225618.1
-
4
257
FQ225678.1
-
4
259
FQ225688.1
-
4
259
FQ225694.1
-
4
261
FQ225722.1
-
4
257
FQ225884.1
-
4
257
FQ225913.1
-
4
259
FQ225914.1
-
4
261
FQ225965.1
-
4
259
FQ226299.1
-
4
241
FQ226354.1
-
4
259
FQ226360.1
-
4
259
FQ226584.1
-
4
259
FQ226599.1
-
4
260
FQ226651.1
-
4
282
FQ226673.1
-
4
240
FQ226731.1
-
4
259
FQ226746.1
-
4
257
FQ227279.1
-
4
259
FQ227628.1
-
4
267
FQ227644.1
-
4
259
FQ227708.1
-
4
259
FQ227725.1
-
4
259
FQ227727.1
-
4
238
FQ227749.1
-
4
259
FQ228032.1
-
4
239
FQ228597.1
-
4
241
FQ229172.1
-
4
259
FQ229199.1
-
4
259
FQ229230.1
-
4
259
FQ229283.1
-
4
241
FQ229299.1
-
4
242
FQ229315.1
-
4
257
FQ229316.1
-
4
259
FQ230029.1
-
4
259
FQ230086.1
-
4
257
FQ230210.1
-
4
260
FQ230381.1
-
4
259
FQ230595.1
-
4
241
FQ230619.1
-
4
259
FQ230667.1
-
4
259
FQ230705.1
-
4
257
FQ230778.1
-
4
259
FQ230864.1
-
4
259
FQ230993.1
-
4
259
FQ231067.1
-
4
259
FQ231166.1
-
4
258
FQ231444.1
-
4
258
FQ231523.1
-
4
259
FQ231543.1
-
4
259
FQ231632.1
-
4
259
FQ231753.1
-
4
256
FQ231815.1
-
4
241
FQ231918.1
-
4
241
FQ231975.1
-
4
241
FQ232042.1
-
4
258
FQ232191.1
-
4
259
FQ232201.1
-
4
259
FQ232223.1
-
4
259
FQ232271.1
-
4
257
FQ232364.1
-
4
261
FQ232445.1
-
4
257
FQ232464.1
-
4
259
FQ232710.1
-
4
257
FQ232838.1
-
4
259
FQ232847.1
-
4
259
FQ232862.1
-
4
257
FQ232869.1
-
4
259
FQ232965.1
-
4
259
FQ232977.1
-
4
259
FQ232986.1
-
4
259
FQ233178.1
-
4
259
FQ233283.1
-
4
238
FQ233669.1
-
4
258
FQ233698.1
-
4
241
FQ233777.1
-
4
257
FQ233810.1
-
4
261
FQ233845.1
-
4
259
FQ233850.1
-
4
260
FQ234037.1
-
4
259
FQ234069.1
-
4
259
FQ234108.1
-
4
259
FQ234199.1
-
4
259
FQ234410.1
-
4
241
FQ234418.1
-
4
259
FQ234523.1
-
4
246
FQ234589.1
-
4
257
FQ234657.1
-
4
241
FQ234685.1
-
4
259
FQ234766.1
-
4
256
FQ235038.1
-
4
259
FQ235049.1
-
4
241
Y13714.1
-
5
225
Back To Top
More Information
Panther Classification:
Molecular Function -
extracellular matrix glycoprotein
Gene Ontology Categories:
Function(s)
Process(es)
ossification
negative regulation of endothelial cell proliferation
signal transduction
heart development
response to gravity
response to lead ion
positive regulation of endothelial cell migration
negative regulation of angiogenesis
regulation of cell morphogenesis
lung development
response to lipopolysaccharide
response to L-ascorbic acid
response to cytokine
wound healing
regulation of cell proliferation
response to peptide hormone
response to ethanol
response to cadmium ion
inner ear development
response to glucocorticoid
response to cAMP
response to calcium ion
bone development
cellular response to growth factor stimulus
calcium ion binding
collagen binding
extracellular matrix binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0