Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYK SIRNA Assays ›
Gene Symbol:
Syk
Gene Name:
spleen tyrosine kinase
Gene Aliases:
p72syk
Chromosome Location:
Chr.17: 12614311 - 12669568 on Build Rnor_6.0
UniGene ID:
Rn.87407
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48428
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Syk
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012758.1
NP_036890.1
10
1473
GenBank mRNA
L20838.1
-
8
1109
U21683.1
-
9
1404
U21684.1
-
10
1473
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001198977, NM_011518, U25685, Z49877, AK143564, and others.
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
angiogenesis
serotonin secretion
lymph vessel development
adaptive immune response
macrophage activation involved in immune response
neutrophil activation involved in immune response
leukocyte activation involved in immune response
serotonin secretion by platelet
protein phosphorylation
inflammatory response
leukocyte cell-cell adhesion
cell surface receptor signaling pathway
enzyme linked receptor protein signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
integrin-mediated signaling pathway
activation of JUN kinase activity
regulation of platelet activation
cell migration
peptidyl-serine phosphorylation
peptidyl-tyrosine phosphorylation
leukotriene biosynthetic process
signal transduction by protein phosphorylation
neutrophil chemotaxis
regulation of superoxide anion generation
positive regulation of cell adhesion mediated by integrin
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
defense response to bacterium
transcription factor import into nucleus
positive regulation of mast cell degranulation
regulation of neutrophil degranulation
beta selection
innate immune response
positive regulation of interleukin-3 biosynthetic process
positive regulation of granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of B cell differentiation
positive regulation of gamma-delta T cell differentiation
positive regulation of bone resorption
positive regulation of alpha-beta T cell differentiation
positive regulation of alpha-beta T cell proliferation
protein autophosphorylation
blood vessel morphogenesis
positive regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
regulation of phagocytosis
regulation of immune response
positive regulation of calcium-mediated signaling
B cell receptor signaling pathway
regulation of sequence-specific DNA binding transcription factor activity
regulation of ERK1 and ERK2 cascade
cellular response to molecule of fungal origin
regulation of arachidonic acid secretion
regulation of platelet aggregation
protein kinase activity
protein serine/threonine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
receptor binding
integrin binding
protein binding
ATP binding
protein kinase binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0