Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DLG1 SIRNA Assays ›
Gene Symbol:
Dlg1
Gene Name:
discs, large homolog 1 (Drosophila)
Gene Aliases:
Dlgh1, SAP97
Chromosome Location:
Chr.11: 72164566 - 72378895 on Build Rnor_6.0
UniGene ID:
Rn.89331
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48450
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dlg1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012788.1
NP_036920.1
10
1392
GenBank mRNA
U14950.1
-
10
1392
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
ureteric bud development
branching involved in ureteric bud morphogenesis
immunological synapse formation
endothelial cell proliferation
lens development in camera-type eye
T cell cytokine production
actin filament organization
cell adhesion
chemical synaptic transmission
nervous system development
protein localization
positive regulation of cell proliferation
single organismal cell-cell adhesion
peristalsis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
membrane raft organization
regulation of myelination
activation of protein kinase activity
regulation of protein localization
T cell activation
negative regulation of T cell proliferation
regulation of membrane potential
amyloid precursor protein metabolic process
receptor clustering
positive regulation of potassium ion transport
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
nucleotide phosphorylation
reproductive structure development
embryonic skeletal system morphogenesis
tissue morphogenesis
smooth muscle tissue development
negative regulation of epithelial cell proliferation
negative regulation of protein kinase B signaling
hard palate development
bicellular tight junction assembly
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
receptor localization to synapse
guanylate kinase activity
protein binding
protein C-terminus binding
potassium channel regulator activity
protein kinase binding
phosphatase binding
PDZ domain binding
mitogen-activated protein kinase kinase binding
protein complex scaffold
ionotropic glutamate receptor binding
ion channel binding
L27 domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0