Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other AHSG SIRNA Assays ›
Gene Symbol:
Ahsg
Gene Name:
alpha-2-HS-glycoprotein
Gene Aliases:
Aa2-066, pp63
Chromosome Location:
Chr.11: 81711269 - 81717594 on Build Rnor_6.0
UniGene ID:
Rn.32083
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48543
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ahsg
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012898.4
NP_037030.1
1
151
GenBank mRNA
AY325170.1
-
787
BC091118.1
-
1
129
7368203
FQ209390.1
-
1
148
FQ209416.1
-
1
150
FQ209417.1
-
1
150
FQ209422.1
-
1
150
FQ209445.1
-
1
150
FQ209451.1
-
1
150
FQ209502.1
-
1
150
FQ209536.1
-
1
150
FQ209545.1
-
1
150
FQ209557.1
-
1
150
FQ209568.1
-
1
150
FQ209581.1
-
1
150
FQ209585.1
-
1
150
FQ209588.1
-
1
150
FQ209604.1
-
1
150
FQ209612.1
-
1
150
FQ209617.1
-
1
150
FQ209654.1
-
1
150
FQ209655.1
-
1
148
FQ209680.1
-
1
150
FQ209720.1
-
1
150
FQ209759.1
-
1
150
FQ209793.1
-
1
150
FQ209828.1
-
1
105
FQ209920.1
-
1
150
FQ209985.1
-
1
150
FQ210030.1
-
1
145
FQ210033.1
-
1
150
FQ210056.1
-
1
148
FQ210082.1
-
1
150
FQ210085.1
-
1
148
FQ210120.1
-
1
149
FQ210139.1
-
1
150
FQ210141.1
-
1
150
FQ210159.1
-
1
150
FQ210164.1
-
1
148
FQ210170.1
-
1
150
FQ210199.1
-
1
150
FQ210228.1
-
1
150
FQ210239.1
-
1
150
FQ210242.1
-
1
147
FQ210249.1
-
1
150
FQ210260.1
-
1
150
FQ210304.1
-
1
150
FQ210382.1
-
1
150
FQ210396.1
-
1
150
FQ210402.1
-
1
150
FQ210405.1
-
1
150
FQ210452.1
-
1
151
FQ210457.1
-
1
150
FQ210498.1
-
1
150
FQ210504.1
-
1
150
FQ210525.1
-
1
149
FQ210562.1
-
1
150
FQ210566.1
-
1
150
FQ210700.1
-
1
150
FQ210709.1
-
1
149
FQ210731.1
-
1
148
FQ210738.1
-
1
150
FQ210739.1
-
1
150
FQ210760.1
-
1
148
FQ210764.1
-
1
150
FQ210871.1
-
1
150
FQ210891.1
-
1
150
FQ210896.1
-
1
148
FQ210953.1
-
1
150
FQ210957.1
-
1
156
FQ210983.1
-
1
150
FQ211020.1
-
1
150
FQ211045.1
-
1
147
FQ211090.1
-
1
150
FQ211092.1
-
1
150
FQ211115.1
-
1
150
FQ211177.1
-
1
150
FQ211179.1
-
1
150
FQ211283.1
-
1
150
FQ211299.1
-
1
150
FQ211301.1
-
1
150
FQ211303.1
-
1
150
FQ211304.1
-
1
150
FQ211368.1
-
1
150
FQ211379.1
-
1
150
FQ211843.1
-
1
150
FQ217075.1
-
1
148
FQ218089.1
-
1
150
FQ218223.1
-
1
149
FQ218236.1
-
1
150
FQ218262.1
-
1
150
FQ218264.1
-
1
150
FQ218269.1
-
1
150
FQ218332.1
-
1
150
FQ218341.1
-
1
150
FQ218342.1
-
1
148
FQ218355.1
-
1
149
FQ218373.1
-
1
150
FQ218390.1
-
1
150
FQ218393.1
-
1
150
FQ218400.1
-
1
150
FQ218424.1
-
1
148
FQ218436.1
-
1
148
FQ218463.1
-
1
148
FQ218464.1
-
1
150
FQ218469.1
-
1
148
FQ218480.1
-
1
150
FQ218487.1
-
1
150
FQ218489.1
-
1
150
FQ218493.1
-
1
150
FQ218500.1
-
1
150
FQ218509.1
-
1
148
FQ218531.1
-
1
150
FQ218533.1
-
1
150
FQ218563.1
-
1
148
FQ218569.1
-
1
148
FQ218570.1
-
1
150
FQ218580.1
-
1
150
FQ218585.1
-
1
150
FQ218608.1
-
1
150
FQ218609.1
-
1
150
FQ218630.1
-
1
146
FQ218634.1
-
1
150
FQ218648.1
-
1
148
FQ218657.1
-
1
150
FQ218695.1
-
1
150
FQ218713.1
-
1
148
FQ218744.1
-
1
132
FQ218765.1
-
1
150
FQ218787.1
-
1
152
FQ218796.1
-
1
150
FQ218800.1
-
1
150
FQ218813.1
-
1
237
FQ218846.1
-
1
150
FQ218858.1
-
1
147
FQ218862.1
-
1
150
FQ218872.1
-
1
150
FQ218877.1
-
1
148
FQ218889.1
-
1
150
FQ218890.1
-
1
150
FQ218892.1
-
1
150
FQ218907.1
-
1
150
FQ218931.1
-
1
149
FQ218948.1
-
1
150
FQ218951.1
-
1
150
FQ218970.1
-
1
148
FQ218974.1
-
1
151
FQ218975.1
-
1
150
FQ218980.1
-
1
150
FQ218981.1
-
1
148
FQ219018.1
-
1
150
FQ219028.1
-
1
150
FQ219033.1
-
1
150
FQ219042.1
-
1
149
FQ219060.1
-
1
150
FQ219061.1
-
1
150
FQ219086.1
-
1
150
FQ219094.1
-
1
150
FQ219099.1
-
1
150
FQ219103.1
-
1
150
FQ219110.1
-
1
150
FQ219121.1
-
1
150
FQ219129.1
-
1
148
FQ219130.1
-
1
146
FQ219141.1
-
1
150
FQ219164.1
-
1
150
FQ219173.1
-
1
150
FQ219194.1
-
1
150
FQ219203.1
-
1
150
FQ219221.1
-
1
150
FQ219232.1
-
1
156
FQ219235.1
-
1
150
FQ219239.1
-
1
150
FQ219251.1
-
1
150
FQ219274.1
-
1
150
FQ219295.1
-
1
150
FQ219300.1
-
1
150
FQ219311.1
-
1
150
FQ219320.1
-
1
150
FQ219326.1
-
1
150
FQ219358.1
-
1
149
FQ219363.1
-
1
152
FQ219364.1
-
1
150
FQ219369.1
-
1
148
FQ219370.1
-
1
150
FQ219377.1
-
1
150
FQ219388.1
-
1
150
FQ219400.1
-
1
150
FQ219406.1
-
1
150
FQ219412.1
-
1
150
FQ219419.1
-
1
148
FQ219486.1
-
1
145
FQ219487.1
-
1
150
FQ219488.1
-
1
150
FQ219536.1
-
1
150
FQ219551.1
-
1
150
FQ219552.1
-
1
148
FQ219558.1
-
1
150
FQ219565.1
-
1
150
FQ219587.1
-
1
148
FQ219606.1
-
1
150
FQ219607.1
-
1
148
FQ219608.1
-
1
148
FQ219617.1
-
1
150
FQ219643.1
-
1
150
FQ219653.1
-
1
149
FQ219660.1
-
1
150
FQ219663.1
-
1
150
FQ219668.1
-
1
151
FQ219672.1
-
1
150
FQ219688.1
-
1
148
FQ219691.1
-
1
150
FQ219726.1
-
1
150
FQ219755.1
-
1
150
FQ219770.1
-
1
150
FQ219772.1
-
1
149
FQ219773.1
-
1
150
FQ219774.1
-
1
150
FQ222367.1
-
1
148
M29758.1
-
1
151
X63446.1
-
1
109
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
ossification
protein complex assembly
acute-phase response
male gonad development
negative regulation of endopeptidase activity
cerebral cortex development
negative regulation of cell growth
negative regulation of bone mineralization
organ regeneration
cellular response to insulin stimulus
negative regulation of phosphorylation
positive regulation of bone resorption
negative regulation of insulin receptor signaling pathway
regulation of inflammatory response
positive regulation of phagocytosis
negative regulation of protein tyrosine kinase activity
cysteine-type endopeptidase inhibitor activity
kinase inhibitor activity
receptor signaling protein tyrosine kinase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline