Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other COL2A1 SIRNA Assays ›
Gene Symbol:
Col2a1
Gene Name:
collagen, type II, alpha 1
Gene Aliases:
CG2A1A, COLLII
Chromosome Location:
Chr.7: 139454945 - 139484098 on Build Rnor_6.0
UniGene ID:
Rn.10124
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48919
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Col2a1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012929.1
NP_037061.1
9
609
XM_006242308.2
XP_006242370.1
10
914
GenBank mRNA
L48440.1
-
9
609
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001844, NM_033150, X16468, BC116449, X16711.
This siRNA also targets the following transcripts in Mouse: NM_031163, NM_001113515, BC082331, BC051383, BC052326.
Panther Classification:
Molecular Function -
extracellular matrix structural protein
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
cartilage condensation
tissue homeostasis
endochondral ossification
chondrocyte differentiation
heart morphogenesis
growth plate cartilage development
proteoglycan metabolic process
central nervous system development
visual perception
sensory perception of sound
response to mechanical stimulus
response to X-ray
regulation of gene expression
collagen fibril organization
notochord development
limb morphogenesis
inner ear morphogenesis
skeletal system morphogenesis
inner ear development
cartilage development
palate development
limb bud formation
embryonic skeletal joint morphogenesis
bone development
cartilage development involved in endochondral bone morphogenesis
cellular response to mechanical stimulus
cellular response to retinoic acid
cellular response to vitamin E
cellular response to nicotine
cellular response to tumor necrosis factor
cellular response to parathyroid hormone stimulus
cellular response to peptide hormone stimulus
otic vesicle development
cellular response to BMP stimulus
response to fibroblast growth factor
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
extracellular matrix structural constituent
identical protein binding
metal ion binding
platelet-derived growth factor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-bznxf:80/100.66.77.153:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline