Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMGB1 SIRNA Assays ›
          Gene Symbol:
          Hmgb1 Hmg1l1 Hmgb1-ps2
          Gene Name:
          high mobility group box 1
          high-mobility group (nonhistone chromosomal) protein 1-like 1
          high mobility group box 1, pseudogene 2
          Gene Aliases:
          Ac2-008, Hmg1
          -
          -
          Chromosome Location:
          Chr.12: 7090927 - 7092190 on Build Rnor_6.0
          Chromosome Location:
          Chr.19: 59984234 - 59985494 on Build Rnor_6.0
          Chromosome Location:
          Chr.17: 30735671 - 30737550 on Build Rnor_6.0
          UniGene ID:
          Rn.144565
          Rn.151172
          Rn.196610
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID 48947

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hmgb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012963.2 NP_037095.1 536
          GenBank mRNA AF275734.1 - 568
          BC061779.1 - 524
          BC081839.1 - 536
          BC088402.1 - 612
          M64986.1 - 601
          Y00463.1 - 506
          Gene Symbol: Hmg1l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC168143.1 - 1 616 9100911
          Gene Symbol: Hmgb1-ps2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC166865.1 - 1 929

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_010439, Z11997, AK050659, BC100312, BC006586, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          eye development
          positive regulation of protein phosphorylation
          positive regulation of mesenchymal cell proliferation
          inflammatory response to antigenic stimulus
          base-excision repair, DNA ligation
          chromatin remodeling
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          chemotaxis
          nervous system development
          circadian rhythm
          negative regulation of DNA replication
          positive regulation of cell proliferation
          response to heat
          response to glucose
          positive regulation of cell death
          positive regulation of neuron projection development
          positive regulation of smooth muscle cell migration
          negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
          lung development
          positive regulation of cell migration
          neuron projection development
          actin cytoskeleton reorganization
          DNA geometric change
          response to lipopolysaccharide
          response to insulin
          positive regulation of myeloid cell apoptotic process
          V(D)J recombination
          response to interferon-gamma
          response to drug
          positive regulation of apoptotic process
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          positive regulation of DNA binding
          positive regulation of myeloid cell differentiation
          positive regulation of myoblast differentiation
          positive regulation of glycogen catabolic process
          positive regulation of protein kinase activity
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          regulation of inflammatory response
          male-specific defense response to bacterium
          induction of positive chemotaxis
          response to glucocorticoid
          myoblast proliferation
          cellular response to interleukin-1
          four-way junction DNA binding
          open form four-way junction DNA binding
          crossed form four-way junction DNA binding
          DNA binding
          bent DNA binding
          chromatin binding
          double-stranded DNA binding
          single-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          cytokine activity
          protein binding
          5S rRNA binding
          transcription factor binding
          heparin binding
          DNA binding, bending
          calcium-dependent protein kinase regulator activity
          protein kinase activator activity
          peptide binding
          poly(A) RNA binding
          protein dimerization activity
          RAGE receptor binding
          glycolipid binding
          repressing transcription factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0