Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APOA2 SIRNA Assays ›
          Gene Symbol:
          Apoa2
          Gene Name:
          apolipoprotein A-II
          Gene Aliases:
          APOAII, Apo-AII, ApoA-II
          Chromosome Location:
          Chr.13: 89596872 - 89598805 on Build Rnor_6.0
          UniGene ID:
          Rn.89304
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 49233

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Apoa2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013112.1 NP_037244.1 2, 3 194
          XM_006250238.2 XP_006250300.1 3, 4 328
          XM_006250239.2 XP_006250301.1 3, 4 334
          GenBank mRNA FQ209753.1 - 3, 4 259
          FQ209821.1 - 3, 4 258
          FQ210214.1 - 3, 4 252
          FQ210245.1 - 3, 4 259
          FQ210325.1 - 3, 4 279
          FQ210458.1 - 3, 4 256
          FQ210462.1 - 3, 4 257
          FQ210537.1 - 2, 3 253
          FQ210542.1 - 2, 3 382
          FQ210648.1 - 3, 4 256
          FQ210885.1 - 3, 4 286
          FQ210979.1 - 3, 4 259
          FQ211006.1 - 3, 4 257
          FQ211265.1 - 3, 4 256
          FQ211383.1 - 3, 4 258
          FQ218158.1 - 3, 4 256
          FQ218273.1 - 3, 4 259
          FQ218326.1 - 3, 4 253
          FQ218756.1 - 3, 4 260
          FQ221741.1 - 3, 4 259
          FQ223965.1 - 3, 4 253
          M28615.1 - 2, 3 163
          X03468.1 - 2, 3 194

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          acute inflammatory response
          negative regulation of cytokine secretion involved in immune response
          protein folding
          phosphatidylcholine biosynthetic process
          lipid transport
          cholesterol metabolic process
          phospholipid catabolic process
          response to glucose
          positive regulation of cholesterol esterification
          negative regulation of very-low-density lipoprotein particle remodeling
          protein oxidation
          peptidyl-methionine modification
          regulation of intestinal cholesterol absorption
          cholesterol transport
          organ regeneration
          regulation of protein stability
          negative regulation of cholesterol transport
          cholesterol efflux
          phospholipid efflux
          triglyceride-rich lipoprotein particle remodeling
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          high-density lipoprotein particle clearance
          lipoprotein metabolic process
          response to drug
          cholesterol homeostasis
          positive regulation of catalytic activity
          response to estrogen
          reverse cholesterol transport
          positive regulation of interleukin-8 biosynthetic process
          diacylglycerol catabolic process
          negative regulation of lipid catabolic process
          positive regulation of lipid catabolic process
          response to glucocorticoid
          negative regulation of lipase activity
          negative regulation of cholesterol import
          negative regulation of cholesterol transporter activity
          lipid transporter activity
          phospholipid binding
          high-density lipoprotein particle binding
          lipid binding
          cholesterol binding
          cholesterol transporter activity
          phosphatidylcholine binding
          apolipoprotein receptor binding
          protein homodimerization activity
          protein heterodimerization activity
          lipase inhibitor activity
          phosphatidylcholine-sterol O-acyltransferase activator activity
          high-density lipoprotein particle receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0