Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TF SIRNA Assays ›
          Gene Symbol:
          Tf Srprb
          Gene Name:
          transferrin
          signal recognition particle receptor, B subunit
          Gene Aliases:
          Tfn, Trf
          Ab2-417, Ba1-667, Cc1-8
          Chromosome Location:
          Chr.8: 111694570 - 111721275 on Build Rnor_6.0
          Chromosome Location:
          Chr.8: 111673479 - 111687426 on Build Rnor_6.0
          UniGene ID:
          Rn.91296
          Rn.138078
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 49748

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Tf
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013110.1 NP_001013128.1 13 1632
          GenBank mRNA AF476964.1 - 13 1663
          AY327504.1 - 13 1601
          BC087021.1 - 13 1632 7132382
          D38380.1 - 13 1631
          FQ209414.1 - 6 596
          FQ209524.1 - 6 691
          FQ209552.1 - 4 411
          FQ209673.1 - 7 799
          FQ209794.1 - 5 484
          FQ210028.1 - 2 217
          FQ210115.1 - 1 112
          FQ210327.1 - 1 88
          FQ210637.1 - 1 31
          FQ210963.1 - 2 156
          FQ211385.1 - 4 408
          FQ213988.1 - 3 317
          FQ218210.1 - 5 462
          FQ219264.1 - 7 735
          FQ219315.1 - 2 273
          FQ219349.1 - 5 465
          FQ219664.1 - 2 126
          M27966.1 - 1 41
          X77158.1 - 13 1609
          Gene Symbol: Srprb
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AY325214.1 - 1599
          AY325230.1 - 1602
          AY325241.1 - 1599

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transfer/carrier protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          retina homeostasis
          iron ion transport
          cellular iron ion homeostasis
          apoptotic process
          acute-phase response
          actin filament organization
          activation of JUN kinase activity
          small GTPase mediated signal transduction
          response to lead ion
          response to organic cyclic compound
          ferric iron transport
          osteoclast differentiation
          positive regulation of myelination
          positive regulation of phosphorylation
          positive regulation of bone resorption
          positive regulation of transcription, DNA-templated
          SMAD protein signal transduction
          ERK1 and ERK2 cascade
          positive regulation of oligodendrocyte progenitor proliferation
          cellular response to cAMP
          cellular response to follicle-stimulating hormone stimulus
          positive regulation of cell motility
          GTP binding
          ferric iron binding
          ferric iron transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0