Search
Search

Ptn| Gene Symbol: Ptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017066.2 | NP_058762.1 | 5 | 553 | ||
| XM_006236290.2 | XP_006236352.1 | 5 | 1937 | |||
| XM_006236294.2 | XP_006236356.1 | 5 | 1533 | |||
| GenBank mRNA | BC062013.1 | - | 5 | 553 | 5621277 | |
| FQ213442.1 | - | 5 | 557 | |||
| FQ223195.1 | - | 5 | 705 | |||
| M55601.1 | - | 5 | 754 | |||