Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS3A SIRNA Assays ›
          Gene Symbol:
          Rps3a
          Gene Name:
          ribosomal protein S3a
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 185440477 - 185444846 on Build Rnor_6.0
          UniGene ID:
          Rn.94935
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 49832

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps3a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017153.1 NP_058849.1 2, 3 184
          GenBank mRNA BC058483.1 - 2, 3 189 6887884
          FQ209727.1 - 2, 3 184
          FQ211699.1 - 2, 3 185
          FQ211708.1 - 2, 3 184
          FQ216850.1 - 2, 3 184
          FQ217809.1 - 2, 3 184
          FQ219020.1 - 2, 3 184
          FQ219404.1 - 2, 3 183
          FQ219522.1 - 2, 3 185
          FQ219543.1 - 2, 3 185
          FQ219867.1 - 2, 3 185
          FQ220002.1 - 2, 3 184
          FQ220027.1 - 2, 3 184
          FQ220037.1 - 2, 3 185
          FQ220045.1 - 2, 3 184
          FQ220135.1 - 2, 3 185
          FQ220260.1 - 2, 3 184
          FQ220261.1 - 2, 3 185
          FQ220317.1 - 2, 3 184
          FQ220504.1 - 2, 3 184
          FQ220577.1 - 2, 3 185
          FQ220731.1 - 2, 3 184
          FQ221185.1 - 2, 3 185
          FQ221431.1 - 2, 3 185
          FQ221460.1 - 2, 3 184
          FQ221744.1 - 2, 3 184
          FQ221931.1 - 2, 3 184
          FQ222056.1 - 2, 3 184
          FQ222736.1 - 2, 3 185
          FQ222773.1 - 2, 3 185
          FQ222846.1 - 2, 3 214
          FQ222869.1 - 2, 3 184
          FQ222876.1 - 2, 3 184
          FQ223535.1 - 2, 3 191
          FQ223540.1 - 2, 3 184
          FQ225080.1 - 2, 3 185
          FQ225145.1 - 2, 3 184
          FQ227696.1 - 2, 3 184
          FQ227805.1 - 2, 3 185
          FQ227878.1 - 2, 3 185
          FQ228312.1 - 2, 3 185
          FQ228317.1 - 2, 3 184
          FQ228376.1 - 2, 3 185
          FQ228558.1 - 2, 3 184
          FQ229234.1 - 2, 3 184
          FQ229381.1 - 2, 3 185
          FQ229520.1 - 2, 3 184
          FQ229870.1 - 2, 3 184
          FQ229960.1 - 2, 3 184
          FQ231162.1 - 2, 3 185
          FQ231673.1 - 2, 3 184
          FQ232329.1 - 2, 3 186
          FQ232836.1 - 2, 3 184
          FQ232928.1 - 2, 3 185
          FQ233768.1 - 2, 3 188
          FQ234311.1 - 2, 3 184
          FQ234537.1 - 2, 3 184
          M84716.1 - 2, 3 178
          X75161.1 - 2, 3 184

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoplasmic translation
          translation
          biological_process
          cell differentiation
          negative regulation of apoptotic process
          molecular_function
          structural constituent of ribosome
          translation initiation factor binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0