Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TGFBR3 SIRNA Assays ›
Gene Symbol:
Tgfbr3
Gene Name:
transforming growth factor, beta receptor III
Gene Aliases:
betaglycan
Chromosome Location:
Chr.14: 3506416 - 3680508 on Build Rnor_6.0
UniGene ID:
Rn.9953
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50296
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tgfbr3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017256.1
NP_058952.1
7
1218
GenBank mRNA
M77809.1
-
7
1218
M80784.1
-
7
1203
Back To Top
More Information
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
blood vessel development
response to hypoxia
in utero embryonic development
blastocyst development
epithelial to mesenchymal transition
liver development
protein complex assembly
protein phosphorylation
immune response
signal transduction
transforming growth factor beta receptor signaling pathway
transforming growth factor beta receptor complex assembly
negative regulation of epithelial cell migration
negative regulation of epithelial to mesenchymal transition
cell growth
BMP signaling pathway
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
organ regeneration
response to follicle-stimulating hormone
response to prostaglandin E
response to luteinizing hormone
intracellular signal transduction
regulation of protein binding
negative regulation of epithelial cell proliferation
positive regulation of NF-kappaB transcription factor activity
negative regulation of cellular component movement
ventricular cardiac muscle tissue morphogenesis
palate development
cardiac muscle cell proliferation
definitive hemopoiesis
cardiac epithelial to mesenchymal transition
definitive erythrocyte differentiation
heart trabecula formation
pathway-restricted SMAD protein phosphorylation
transforming growth factor beta-activated receptor activity
type II transforming growth factor beta receptor binding
transforming growth factor beta receptor binding
protein binding
glycosaminoglycan binding
heparin binding
coreceptor activity
fibroblast growth factor binding
PDZ domain binding
SMAD binding
activin binding
transforming growth factor beta binding
transforming growth factor beta receptor activity, type III
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0