Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHD7 SIRNA Assays ›
Gene Symbol:
Chd7
Gene Name:
chromodomain helicase DNA binding protein 7
Gene Aliases:
A730019I05Rik, Cycn, Cyn, Dz, Edy, Flo, GENA 47, GENA 60, Gena 52, Lda, Mt, Obt, Todo, WBE1, Whi, metis
Chromosome Location:
Chr.4: 8690406 - 8868449 on Build GRCm38
UniGene ID:
Mm.138792
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
503575
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Chd7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001277149.1
NP_001264078.1
12
3554
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
skeletal system development
blood vessel development
in utero embryonic development
heart morphogenesis
transcription, DNA-templated
regulation of transcription, DNA-templated
rRNA processing
central nervous system development
adult heart development
sensory perception of sound
locomotory behavior
adult walking behavior
blood circulation
chromatin modification
cranial nerve development
olfactory nerve development
olfactory bulb development
T cell differentiation
female genitalia development
embryonic hindlimb morphogenesis
positive regulation of multicellular organism growth
olfactory behavior
ear morphogenesis
inner ear morphogenesis
camera-type eye development
nose development
semicircular canal morphogenesis
genitalia development
artery morphogenesis
regulation of neurogenesis
cognition
palate development
retina development in camera-type eye
regulation of growth hormone secretion
limb development
face development
epithelium development
nucleotide binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
DNA binding
chromatin binding
helicase activity
protein binding
ATP binding
hydrolase activity
hydrolase activity, acting on acid anhydrides
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0