Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACTA1 SIRNA Assays ›
Gene Symbol:
Acta1
Gene Name:
actin, alpha 1, skeletal muscle
Gene Aliases:
-
Chromosome Location:
Chr.19: 56674072 - 56677084 on Build Rnor_6.0
UniGene ID:
Rn.82732
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50645
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acta1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019212.2
NP_062085.1
6
950
GenBank mRNA
BC061974.1
-
6
950
5623909
FQ214583.1
-
6
960
FQ214585.1
-
6
961
FQ214598.1
-
6
961
FQ214600.1
-
6
960
FQ214603.1
-
6
963
FQ214610.1
-
4
504
FQ214640.1
-
6
959
FQ214659.1
-
6
961
FQ214676.1
-
6
960
FQ214680.1
-
6
960
FQ214697.1
-
6
965
FQ214734.1
-
6
960
FQ214743.1
-
6
959
FQ214756.1
-
6
961
FQ214761.1
-
6
958
FQ214779.1
-
6
958
FQ214785.1
-
6
958
FQ214807.1
-
6
959
FQ214822.1
-
6
960
FQ214831.1
-
6
960
FQ214832.1
-
6
958
FQ214839.1
-
6
962
FQ214841.1
-
6
959
FQ214842.1
-
6
960
FQ214857.1
-
6
963
FQ214858.1
-
6
959
FQ214859.1
-
6
959
FQ214860.1
-
6
961
FQ214862.1
-
6
959
FQ214866.1
-
6
961
FQ214883.1
-
6
958
FQ214904.1
-
6
959
FQ214905.1
-
6
960
FQ214916.1
-
6
959
FQ214921.1
-
6
960
FQ214936.1
-
6
960
FQ214948.1
-
6
959
FQ214949.1
-
6
960
FQ214957.1
-
6
959
FQ214969.1
-
6
959
FQ214975.1
-
6
962
FQ214976.1
-
6
959
FQ214978.1
-
6
959
FQ214987.1
-
6
959
FQ214990.1
-
6
959
FQ214993.1
-
6
959
FQ215012.1
-
6
960
FQ215025.1
-
6
961
FQ215038.1
-
6
958
FQ215044.1
-
6
959
FQ215058.1
-
6
960
FQ215089.1
-
6
960
FQ215112.1
-
6
959
FQ215128.1
-
6
961
FQ215136.1
-
6
959
FQ215149.1
-
6
961
FQ215153.1
-
6
959
FQ215158.1
-
6
957
FQ215173.1
-
6
960
FQ215185.1
-
6
959
FQ215204.1
-
6
957
FQ215215.1
-
6
960
FQ215225.1
-
6
960
FQ215241.1
-
6
960
FQ215249.1
-
6
960
FQ215253.1
-
6
960
FQ215256.1
-
6
959
FQ215275.1
-
6
959
FQ215277.1
-
6
960
FQ215280.1
-
6
960
FQ215282.1
-
6
959
FQ215298.1
-
6
960
FQ215324.1
-
6
959
FQ215333.1
-
6
959
FQ215342.1
-
6
959
FQ215346.1
-
6
959
FQ215366.1
-
6
960
FQ215380.1
-
6
961
FQ215381.1
-
6
960
FQ215393.1
-
6
959
FQ215396.1
-
6
959
FQ215398.1
-
6
959
FQ215402.1
-
6
959
FQ215403.1
-
6
960
FQ215405.1
-
6
958
FQ215414.1
-
6
961
FQ215424.1
-
6
960
FQ215431.1
-
6
960
FQ215433.1
-
6
958
FQ215434.1
-
6
961
FQ215438.1
-
6
959
FQ215449.1
-
6
962
FQ215454.1
-
6
960
FQ215464.1
-
6
959
FQ215465.1
-
6
959
FQ215468.1
-
6
959
FQ215470.1
-
6
959
FQ215474.1
-
6
960
FQ215483.1
-
6
959
FQ215488.1
-
6
959
FQ215518.1
-
6
968
FQ215523.1
-
6
959
FQ215537.1
-
6
961
FQ215554.1
-
6
959
FQ215567.1
-
6
961
FQ215572.1
-
6
960
FQ215593.1
-
6
959
FQ215618.1
-
6
960
FQ215628.1
-
6
962
FQ215631.1
-
4
573
FQ215655.1
-
6
957
FQ215659.1
-
6
958
FQ215673.1
-
6
959
FQ215674.1
-
6
959
FQ215690.1
-
6
960
FQ215692.1
-
6
960
FQ215785.1
-
6
959
FQ215790.1
-
6
961
FQ215799.1
-
6
958
FQ215809.1
-
6
959
FQ215813.1
-
6
959
FQ215839.1
-
6
959
FQ215840.1
-
6
960
FQ215851.1
-
6
959
FQ215852.1
-
6
957
FQ215864.1
-
6
961
FQ215866.1
-
6
960
FQ215888.1
-
6
958
FQ215893.1
-
6
962
FQ215896.1
-
6
959
FQ215907.1
-
6
962
FQ215924.1
-
4
651
FQ215950.1
-
6
961
FQ215964.1
-
6
959
FQ215972.1
-
6
961
FQ216016.1
-
6
959
FQ216035.1
-
6
959
FQ216043.1
-
6
959
FQ216050.1
-
6
959
FQ216066.1
-
6
957
FQ216138.1
-
6
960
FQ216168.1
-
6
958
FQ216196.1
-
6
960
FQ216224.1
-
6
960
FQ216246.1
-
6
960
FQ216276.1
-
6
959
FQ216278.1
-
6
963
FQ216287.1
-
6
959
FQ216291.1
-
6
960
FQ216293.1
-
6
960
FQ216305.1
-
6
960
FQ216329.1
-
6
959
FQ216333.1
-
6
960
FQ216350.1
-
6
959
FQ216352.1
-
6
959
FQ216360.1
-
6
960
FQ216369.1
-
6
962
FQ216403.1
-
6
959
FQ216430.1
-
6
960
FQ216509.1
-
6
957
FQ216586.1
-
6
959
FQ216603.1
-
6
961
FQ216606.1
-
6
959
FQ216607.1
-
6
955
FQ216635.1
-
6
957
FQ216639.1
-
6
959
FQ216644.1
-
6
960
FQ216664.1
-
6
957
FQ216675.1
-
6
959
FQ216676.1
-
6
959
FQ216692.1
-
6
960
FQ216703.1
-
6
959
FQ216713.1
-
6
959
FQ216721.1
-
6
959
FQ216723.1
-
6
960
FQ216725.1
-
6
955
FQ216735.1
-
2
199
FQ216739.1
-
6
959
FQ216762.1
-
6
957
FQ216767.1
-
6
959
FQ216776.1
-
6
960
FQ216777.1
-
6
959
FQ216779.1
-
6
961
FQ216793.1
-
6
959
FQ216799.1
-
6
957
FQ216801.1
-
6
959
FQ216814.1
-
6
954
FQ216819.1
-
6
957
FQ216916.1
-
6
961
FQ216918.1
-
4
602
FQ216961.1
-
6
960
FQ216994.1
-
6
959
FQ216999.1
-
2
158
FQ217061.1
-
6
961
FQ217073.1
-
6
962
FQ217091.1
-
6
959
FQ217114.1
-
6
961
FQ217122.1
-
6
959
FQ217167.1
-
6
959
FQ217173.1
-
6
959
FQ217188.1
-
6
959
FQ217194.1
-
6
959
FQ217266.1
-
1
86
FQ217273.1
-
6
958
FQ217280.1
-
6
1079
FQ217327.1
-
6
960
FQ217329.1
-
6
959
FQ217347.1
-
6
958
FQ217351.1
-
1
42
FQ217353.1
-
6
959
FQ217367.1
-
6
959
FQ217397.1
-
6
959
FQ217441.1
-
6
959
FQ217443.1
-
6
961
FQ217475.1
-
6
960
FQ217525.1
-
6
959
FQ217535.1
-
6
959
FQ217572.1
-
6
959
FQ217593.1
-
6
959
FQ217602.1
-
6
961
FQ217615.1
-
6
959
FQ217629.1
-
6
961
FQ217641.1
-
6
956
FQ217668.1
-
2
208
FQ217762.1
-
6
959
FQ217826.1
-
6
959
FQ217855.1
-
1
34
FQ217884.1
-
6
958
FQ217893.1
-
6
960
FQ217905.1
-
6
959
FQ217908.1
-
6
961
FQ217919.1
-
6
959
FQ217937.1
-
6
959
FQ217969.1
-
6
961
FQ218005.1
-
3
359
FQ218019.1
-
6
960
FQ218034.1
-
6
959
FQ223563.1
-
6
960
FQ223686.1
-
6
959
FQ223702.1
-
6
960
FQ223727.1
-
4
657
FQ223778.1
-
4
473
FQ223898.1
-
6
960
FQ223914.1
-
6
959
FQ223926.1
-
6
959
FQ223929.1
-
6
958
FQ224009.1
-
6
959
FQ224082.1
-
6
959
FQ224141.1
-
6
960
FQ224167.1
-
6
961
FQ224359.1
-
6
960
FQ224377.1
-
6
959
FQ224419.1
-
6
959
FQ224552.1
-
6
960
FQ224777.1
-
6
960
FQ224840.1
-
6
957
FQ224842.1
-
6
959
FQ224869.1
-
6
960
FQ224886.1
-
6
958
FQ224890.1
-
6
957
FQ224902.1
-
6
960
FQ224929.1
-
6
957
FQ224936.1
-
6
960
FQ224942.1
-
6
962
FQ224946.1
-
4
487
FQ224948.1
-
6
964
FQ224962.1
-
6
961
FQ224971.1
-
6
959
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: BC014877.
Panther Classification:
Molecular Function -
actin and actin related protein
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
response to mechanical stimulus
response to extracellular stimulus
response to lithium ion
cell growth
skeletal muscle thin filament assembly
skeletal muscle fiber adaptation
response to steroid hormone
skeletal muscle fiber development
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0