Search
Search

| Gene Symbol: Pdf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026513.2 | NP_080789.2 | 2 | 1159 | ||
| GenBank mRNA | AK008820.1 | - | 2 | 799 | ||
| AK011473.1 | - | 2 | 712 | |||
| AK019363.1 | - | 1 | 42 | |||
| Gene Symbol: Cog8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC023861.1 | - | 1166 | |||
| BC025453.1 | - | 541 | ||||
| BC094654.1 | - | 4075 | 6853777 | |||