Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACTA1 SIRNA Assays ›
          Gene Symbol:
          Acta1
          Gene Name:
          actin, alpha 1, skeletal muscle
          Gene Aliases:
          -
          Chromosome Location:
          Chr.19: 56674072 - 56677084 on Build Rnor_6.0
          UniGene ID:
          Rn.82732
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 50738

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Acta1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019212.2 NP_062085.1 6 1022
          GenBank mRNA BC061974.1 - 6 1022 5623909
          FQ214583.1 - 6 1032
          FQ214598.1 - 6 1033
          FQ214603.1 - 6 1035
          FQ214610.1 - 4 576
          FQ214623.1 - 6 1030
          FQ214659.1 - 6 1033
          FQ214676.1 - 6 1032
          FQ214680.1 - 6 1032
          FQ214697.1 - 6 1037
          FQ214756.1 - 6 1033
          FQ214761.1 - 6 1030
          FQ214779.1 - 6 1030
          FQ214785.1 - 6 1030
          FQ214786.1 - 6 1034
          FQ214866.1 - 6 1033
          FQ214883.1 - 6 1030
          FQ214904.1 - 6 1031
          FQ214905.1 - 6 1032
          FQ214936.1 - 6 1032
          FQ214978.1 - 6 1031
          FQ214987.1 - 6 1031
          FQ214993.1 - 6 1031
          FQ215025.1 - 6 1033
          FQ215038.1 - 6 1030
          FQ215044.1 - 6 1031
          FQ215089.1 - 6 1032
          FQ215112.1 - 6 1031
          FQ215136.1 - 6 1031
          FQ215173.1 - 6 1032
          FQ215185.1 - 6 1031
          FQ215204.1 - 6 1029
          FQ215212.1 - 6 1033
          FQ215241.1 - 6 1032
          FQ215249.1 - 6 1032
          FQ215256.1 - 6 1031
          FQ215275.1 - 6 1031
          FQ215282.1 - 6 1031
          FQ215333.1 - 6 1031
          FQ215342.1 - 6 1031
          FQ215346.1 - 6 1031
          FQ215381.1 - 6 1032
          FQ215398.1 - 6 1031
          FQ215402.1 - 6 1031
          FQ215403.1 - 6 1032
          FQ215405.1 - 6 1030
          FQ215424.1 - 6 1032
          FQ215431.1 - 6 1032
          FQ215434.1 - 6 1033
          FQ215449.1 - 6 1034
          FQ215454.1 - 6 1032
          FQ215470.1 - 6 1031
          FQ215518.1 - 6 1040
          FQ215537.1 - 6 1033
          FQ215554.1 - 6 1031
          FQ215567.1 - 6 1033
          FQ215572.1 - 6 1032
          FQ215593.1 - 6 1031
          FQ215618.1 - 6 1032
          FQ215628.1 - 6 1034
          FQ215631.1 - 4 645
          FQ215655.1 - 6 1029
          FQ215659.1 - 6 1030
          FQ215673.1 - 6 1031
          FQ215674.1 - 6 1031
          FQ215799.1 - 6 1030
          FQ215809.1 - 6 1031
          FQ215813.1 - 6 1031
          FQ215836.1 - 6 1031
          FQ215839.1 - 6 1031
          FQ215841.1 - 6 1032
          FQ215888.1 - 6 1030
          FQ215893.1 - 6 1034
          FQ215896.1 - 6 1031
          FQ215924.1 - 4 723
          FQ215950.1 - 6 1033
          FQ215972.1 - 6 1033
          FQ216016.1 - 6 1031
          FQ216035.1 - 6 1031
          FQ216043.1 - 6 1031
          FQ216050.1 - 6 1031
          FQ216066.1 - 6 1029
          FQ216138.1 - 6 1032
          FQ216139.1 - 6 1031
          FQ216168.1 - 6 1030
          FQ216196.1 - 6 1032
          FQ216224.1 - 6 1032
          FQ216246.1 - 6 1032
          FQ216276.1 - 6 1031
          FQ216278.1 - 6 1035
          FQ216305.1 - 6 1032
          FQ216333.1 - 6 1032
          FQ216352.1 - 6 1031
          FQ216360.1 - 6 1032
          FQ216369.1 - 6 1034
          FQ216403.1 - 6 1031
          FQ216509.1 - 6 1029
          FQ216586.1 - 6 1031
          FQ216606.1 - 6 1031
          FQ216610.1 - 6 1029
          FQ216620.1 - 6 1032
          FQ216639.1 - 6 1031
          FQ216644.1 - 6 1032
          FQ216676.1 - 6 1031
          FQ216713.1 - 6 1031
          FQ216723.1 - 6 1032
          FQ216725.1 - 6 1027
          FQ216735.1 - 2 271
          FQ216739.1 - 6 1031
          FQ216767.1 - 6 1031
          FQ216776.1 - 6 1032
          FQ216777.1 - 6 1031
          FQ216779.1 - 6 1033
          FQ216793.1 - 6 1031
          FQ216801.1 - 6 1031
          FQ216819.1 - 6 1029
          FQ216916.1 - 6 1033
          FQ216918.1 - 4 674
          FQ216961.1 - 6 1032
          FQ216999.1 - 2 230
          FQ217091.1 - 6 1031
          FQ217173.1 - 6 1031
          FQ217188.1 - 6 1031
          FQ217194.1 - 6 1031
          FQ217266.1 - 1 158
          FQ217280.1 - 6 1151
          FQ217329.1 - 6 1031
          FQ217347.1 - 6 1030
          FQ217351.1 - 1 114
          FQ217353.1 - 6 1031
          FQ217441.1 - 6 1031
          FQ217443.1 - 6 1033
          FQ217535.1 - 6 1031
          FQ217641.1 - 6 1028
          FQ217668.1 - 2 280
          FQ217762.1 - 6 1031
          FQ217826.1 - 6 1031
          FQ217855.1 - 1 106
          FQ217884.1 - 6 1030
          FQ217893.1 - 6 1032
          FQ217905.1 - 6 1031
          FQ217937.1 - 6 1031
          FQ218005.1 - 3 431
          FQ223563.1 - 6 1032
          FQ223686.1 - 6 1031
          FQ223702.1 - 6 1032
          FQ223727.1 - 4 729
          FQ223778.1 - 4 545
          FQ223898.1 - 6 1032
          FQ223914.1 - 6 1031
          FQ223926.1 - 6 1031
          FQ223929.1 - 6 1030
          FQ224009.1 - 6 1031
          FQ224082.1 - 6 1031
          FQ224141.1 - 6 1032
          FQ224359.1 - 6 1032
          FQ224377.1 - 6 1031
          FQ224552.1 - 6 1032
          FQ224691.1 - 6 1029
          FQ224840.1 - 6 1029
          FQ224862.1 - 6 1031
          FQ224869.1 - 6 1032
          FQ224886.1 - 6 1030
          FQ224890.1 - 6 1029
          FQ224902.1 - 6 1032
          FQ224936.1 - 6 1032
          FQ224942.1 - 6 1034
          FQ224946.1 - 4 559
          FQ224948.1 - 6 1036
          FQ224962.1 - 6 1033
          FQ224971.1 - 6 1031

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_001272041, NM_009606, M12866, BC014877, X03766, and others.

          Panther Classification:

          Molecular Function -

          actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          response to mechanical stimulus
          response to extracellular stimulus
          response to lithium ion
          cell growth
          skeletal muscle thin filament assembly
          skeletal muscle fiber adaptation
          response to steroid hormone
          skeletal muscle fiber development
          ATP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0