Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GREM1 SIRNA Assays ›
Gene Symbol:
Grem1
Gene Name:
gremlin 1
Gene Aliases:
Cktsf1b1, drm
Chromosome Location:
Chr.3: 105203309 - 105214989 on Build Rnor_6.0
UniGene ID:
Rn.42929
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50792
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Grem1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_019282.3
NP_062155.1
2
261
GenBank mRNA
Y10019.1
-
2
260
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
angiogenesis
branching involved in ureteric bud morphogenesis
cell migration involved in sprouting angiogenesis
positive regulation of receptor internalization
negative regulation of leukocyte chemotaxis
positive regulation of transcription from RNA polymerase II promoter involved in myocardial precursor cell differentiation
mesenchymal to epithelial transition involved in metanephros morphogenesis
apoptotic process
signal transduction
activation of transmembrane receptor protein tyrosine kinase activity
cell-cell signaling
positive regulation of cell proliferation
organ morphogenesis
proximal/distal pattern formation
regulation of epithelial to mesenchymal transition
collagen fibril organization
negative regulation of cell growth
embryonic limb morphogenesis
negative regulation of bone mineralization
negative regulation of BMP signaling pathway
negative regulation of chondrocyte differentiation
regulation of stress-activated MAPK cascade
negative regulation of osteoblast proliferation
positive regulation of NF-kappaB import into nucleus
negative regulation of apoptotic process
endothelial cell migration
positive regulation of angiogenesis
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of bone remodeling
determination of dorsal identity
positive regulation of NF-kappaB transcription factor activity
regulation of focal adhesion assembly
positive regulation of telomerase activity
limb development
negative regulation of pathway-restricted SMAD protein phosphorylation
ureteric bud formation
positive regulation of protein tyrosine kinase activity
signal transduction by p53 class mediator
negative regulation of monocyte chemotaxis
negative regulation of canonical Wnt signaling pathway
positive regulation of branching involved in ureteric bud morphogenesis
negative regulation of branching involved in ureteric bud morphogenesis
negative regulation of osteoclast proliferation
positive regulation of peptidyl-tyrosine autophosphorylation
negative regulation of bone trabecula formation
negative regulation of bone mineralization involved in bone maturation
positive regulation of receptor activity
positive regulation of cardiac muscle cell differentiation
cytokine activity
protein binding
transmembrane receptor protein tyrosine kinase activator activity
vascular endothelial growth factor receptor 2 binding
receptor agonist activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0