Search
Search

Clasp1| Gene Symbol: Clasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293300.1 | NP_001280229.1 | 7 | 1332 | ||
| NM_001293301.1 | NP_001280230.1 | 7 | 1332 | |||
| NM_029709.2 | NP_083985.2 | 7 | 1332 | |||
| GenBank mRNA | AJ276962.1 | - | 7 | 932 | ||
| AK031181.1 | - | 7 | 934 | |||
| AK042402.1 | - | 7 | 1336 | |||
| AK080782.1 | - | 7 | 1048 | |||
| BC094432.1 | - | 6 | 873 | 30674963 | ||
| BC157938.1 | - | 7 | 934 | |||
| BC158064.1 | - | 7 | 935 | 9007363 | ||