Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSK SIRNA Assays ›
          Gene Symbol:
          CSK
          Gene Name:
          c-src tyrosine kinase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 74782064 - 74803198 on Build GRCh38
          UniGene ID:
          Hs.77793
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 512
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001127190.1 NP_001120662.1 14 1894
          NM_004383.2 NP_004374.1 13 2413
          XM_005254165.4 XP_005254222.1 13 1863
          XM_017021925.1 XP_016877414.1 13 2454
          GenBank mRNA AK127672.1 - 15 2143
          AK290531.1 - 14 1885
          AY007162.1 - 11 1643
          BC104847.1 - 13 1871 8143850
          BC104875.1 - 13 1871 8143878
          BC106073.1 - 13 2413 6526810
          X59932.1 - 13 1844
          X60114.1 - 13 2123

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001030039, XM_006243163, XM_006243164, X58631, FQ227277, and others.
          This siRNA also targets the following transcripts in Mouse: AK194012, AK219690, AK013057, AK170815, BC095962, and others.

          Matching Taqman Gene Expression Assay:


          Hs00177843_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of cytokine production
          adaptive immune response
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          epidermal growth factor receptor signaling pathway
          central nervous system development
          brain development
          negative regulation of cell proliferation
          negative regulation of low-density lipoprotein particle clearance
          cell migration
          cell differentiation
          T cell costimulation
          negative regulation of interleukin-6 production
          negative regulation of kinase activity
          adherens junction organization
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          negative regulation of Golgi to plasma membrane protein transport
          positive regulation of MAP kinase activity
          innate immune response
          negative regulation of bone resorption
          oligodendrocyte differentiation
          negative regulation of phagocytosis
          T cell receptor signaling pathway
          regulation of Fc receptor mediated stimulatory signaling pathway
          negative regulation of ERK1 and ERK2 cascade
          cellular response to peptide hormone stimulus
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          receptor binding
          protein binding
          ATP binding
          protein C-terminus binding
          kinase activity
          protein phosphatase binding
          protein kinase A catalytic subunit binding
          identical protein binding
          metal ion binding
          proline-rich region binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0