Search
Search

Nucb2| Gene Symbol: Nucb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021663.2 | NP_067695.1 | 3, 4 | 352 | ||
| XM_006230095.2 | XP_006230157.1 | 2, 3 | 260 | |||
| GenBank mRNA | AF238223.1 | - | 1, 2 | 139 | ||
| AF250142.1 | - | 2, 3 | 239 | |||
| BC061778.1 | - | 3, 4 | 352 | 5621784 | ||