Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPLP0 SIRNA Assays ›
          Gene Symbol:
          Rplp0 RGD1564469
          Gene Name:
          ribosomal protein, large, P0
          similar to Acidic ribosomal phosphoprotein P0
          Gene Aliases:
          Arbp
          -
          Chromosome Location:
          Chr.12: 46791528 - 46794797 on Build Rnor_6.0
          Chromosome Location:
          Chr.2: 206902779 - 206903920 on Build Rnor_6.0
          UniGene ID:
          Rn.1079
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 51781

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rplp0
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_022402.2 NP_071797.1 3 357
          GenBank mRNA BC062028.1 - 2 326 5623881
          DN931898.1 - 2 375
          FQ209537.1 - 3 357
          FQ209603.1 - 3 357
          FQ209620.1 - 2 342
          FQ209649.1 - 3 357
          FQ209757.1 - 3 358
          FQ210412.1 - 3 357
          FQ211152.1 - 3 357
          FQ211263.1 - 3 356
          FQ212554.1 - 3 357
          FQ212743.1 - 3 357
          FQ212875.1 - 3 357
          FQ212927.1 - 3 358
          FQ213085.1 - 3 357
          FQ213427.1 - 3 357
          FQ213510.1 - 3 357
          FQ213712.1 - 3 357
          FQ213847.1 - 3 358
          FQ214087.1 - 3 357
          FQ214354.1 - 3 357
          FQ214515.1 - 3 357
          FQ214553.1 - 3 358
          FQ214850.1 - 3 357
          FQ214997.1 - 3 357
          FQ215007.1 - 3 357
          FQ215548.1 - 3 359
          FQ215662.1 - 3 357
          FQ215682.1 - 3 357
          FQ215870.1 - 3 357
          FQ216120.1 - 3 358
          FQ216296.1 - 3 357
          FQ216389.1 - 3 358
          FQ216406.1 - 3 360
          FQ216618.1 - 3 357
          FQ216655.1 - 3 358
          FQ216733.1 - 3 359
          FQ216774.1 - 3 357
          FQ216813.1 - 3 356
          FQ216851.1 - 3 357
          FQ216913.1 - 3 358
          FQ217406.1 - 3 357
          FQ218561.1 - 3 357
          FQ218832.1 - 3 357
          FQ218943.1 - 3 357
          FQ219025.1 - 3 360
          FQ219184.1 - 3 361
          FQ219389.1 - 3 357
          FQ219457.1 - 3 357
          FQ219678.1 - 3 357
          FQ219815.1 - 3 357
          FQ219817.1 - 3 357
          FQ219837.1 - 3 357
          FQ219855.1 - 3 357
          FQ219923.1 - 3 357
          FQ220008.1 - 3 357
          FQ220051.1 - 3 357
          FQ220077.1 - 3 357
          FQ220115.1 - 3 357
          FQ220181.1 - 3 357
          FQ220201.1 - 3 355
          FQ220213.1 - 3 357
          FQ220247.1 - 3 358
          FQ220282.1 - 3 359
          FQ220284.1 - 3 357
          FQ220302.1 - 3 357
          FQ220338.1 - 3 357
          FQ220345.1 - 3 357
          FQ220370.1 - 3 357
          FQ220406.1 - 3 357
          FQ220413.1 - 3 357
          FQ220420.1 - 3 358
          FQ220433.1 - 3 354
          FQ220444.1 - 3 357
          FQ220445.1 - 3 353
          FQ220500.1 - 3 357
          FQ220514.1 - 3 357
          FQ220527.1 - 3 357
          FQ220533.1 - 3 389
          FQ220542.1 - 3 357
          FQ220545.1 - 3 357
          FQ220634.1 - 3 357
          FQ220654.1 - 3 357
          FQ220663.1 - 3 357
          FQ220684.1 - 3 358
          FQ220690.1 - 3 359
          FQ220769.1 - 3 357
          FQ220787.1 - 3 357
          FQ220801.1 - 3 357
          FQ220833.1 - 3 357
          FQ220838.1 - 3 357
          FQ220888.1 - 3 357
          FQ220932.1 - 3 358
          FQ221146.1 - 3 357
          FQ221380.1 - 3 357
          FQ221533.1 - 3 359
          FQ221556.1 - 3 355
          FQ222171.1 - 3 357
          FQ222181.1 - 3 357
          FQ222327.1 - 3 357
          FQ222772.1 - 3 357
          FQ222906.1 - 3 357
          FQ223121.1 - 3 357
          FQ223487.1 - 3 357
          FQ224547.1 - 3 357
          FQ224834.1 - 3 357
          FQ224846.1 - 3 357
          FQ225049.1 - 3 357
          FQ225170.1 - 3 357
          FQ225608.1 - 3 357
          FQ225654.1 - 3 359
          FQ225860.1 - 3 359
          FQ225886.1 - 3 357
          FQ226306.1 - 3 359
          FQ227381.1 - 3 357
          FQ227522.1 - 3 352
          FQ227719.1 - 3 357
          FQ227837.1 - 3 357
          FQ227937.1 - 3 357
          FQ227941.1 - 3 357
          FQ227942.1 - 3 477
          FQ228548.1 - 3 357
          FQ228995.1 - 3 357
          FQ228996.1 - 3 357
          FQ229139.1 - 3 359
          FQ229159.1 - 3 357
          FQ229164.1 - 3 358
          FQ229208.1 - 3 357
          FQ229241.1 - 3 357
          FQ229244.1 - 3 357
          FQ229248.1 - 1 237
          FQ229272.1 - 3 357
          FQ229273.1 - 3 358
          FQ229303.1 - 3 357
          FQ229337.1 - 3 357
          FQ229352.1 - 3 357
          FQ229365.1 - 3 357
          FQ229397.1 - 3 357
          FQ229408.1 - 3 358
          FQ229433.1 - 3 357
          FQ229448.1 - 3 357
          FQ229521.1 - 3 357
          FQ229547.1 - 3 357
          FQ229554.1 - 3 357
          FQ229566.1 - 3 353
          FQ229610.1 - 3 357
          FQ229626.1 - 3 354
          FQ229726.1 - 3 357
          FQ229740.1 - 3 357
          FQ229742.1 - 3 357
          FQ229776.1 - 3 357
          FQ229808.1 - 3 357
          FQ229815.1 - 3 357
          FQ229822.1 - 3 357
          FQ229830.1 - 3 359
          FQ229883.1 - 3 359
          FQ229901.1 - 3 357
          FQ229932.1 - 1 255
          FQ229952.1 - 3 357
          FQ229974.1 - 3 357
          FQ230055.1 - 3 356
          FQ230127.1 - 3 358
          FQ230267.1 - 3 358
          FQ230943.1 - 3 357
          FQ231445.1 - 3 359
          FQ231525.1 - 3 357
          FQ231591.1 - 3 359
          FQ231710.1 - 3 358
          FQ231733.1 - 3 358
          FQ231897.1 - 3 359
          FQ231967.1 - 3 357
          FQ232029.1 - 3 358
          FQ232154.1 - 3 357
          FQ232165.1 - 3 357
          FQ232310.1 - 3 356
          FQ232319.1 - 3 358
          FQ232697.1 - 3 359
          FQ232951.1 - 3 357
          FQ233152.1 - 3 356
          FQ233377.1 - 3 357
          FQ233473.1 - 3 357
          FQ233732.1 - 3 357
          FQ233737.1 - 3 357
          FQ233808.1 - 3 357
          FQ234195.1 - 3 357
          FQ234341.1 - 3 357
          FQ234613.1 - 3 357
          FQ234733.1 - 3 357
          FQ234863.1 - 3 357
          FQ235063.1 - 3 357
          FQ235079.1 - 3 357
          FQ235122.1 - 3 357
          FQ235259.1 - 3 357
          X15096.1 - 3 357
          Z29530.1 - 2 313
          Gene Symbol: RGD1564469
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_227546.7 XP_227546.4 2 356

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translational elongation
          ribosome biogenesis
          cellular response to interleukin-4
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4jtmq:80/100.66.78.38:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0