Search
Search

Crip2| Gene Symbol: Crip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022501.1 | NP_071946.1 | 6, 7 | 623 | ||
| XM_008764943.1 | XP_008763165.1 | 6, 7 | 887 | |||
| GenBank mRNA | BC061774.1 | - | 6, 7 | 592 | 5621903 | |
| D17512.1 | - | 6, 7 | 623 | |||
| FQ226314.1 | - | 6, 7 | 631 | |||
| FQ227529.1 | - | 6, 7 | 633 | |||
| FQ230331.1 | - | 6, 7 | 633 | |||
| FQ233584.1 | - | 6, 7 | 637 | |||