Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPLP0 SIRNA Assays ›
Gene Symbol:
Rplp0
Gene Name:
ribosomal protein, large, P0
Gene Aliases:
Arbp
Chromosome Location:
Chr.12: 46791528 - 46794797 on Build Rnor_6.0
UniGene ID:
Rn.1079
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
51948
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Rplp0
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022402.2
NP_071797.1
7
972
GenBank mRNA
BC062028.1
-
6
941
5623881
FQ209537.1
-
7
972
FQ209603.1
-
7
972
FQ209620.1
-
6
957
FQ209649.1
-
7
972
FQ209757.1
-
7
973
FQ210253.1
-
7
972
FQ210412.1
-
7
972
FQ211152.1
-
7
972
FQ211263.1
-
7
971
FQ212554.1
-
7
972
FQ212743.1
-
7
972
FQ212875.1
-
7
972
FQ212927.1
-
7
973
FQ213085.1
-
7
972
FQ213427.1
-
7
972
FQ213510.1
-
7
972
FQ213712.1
-
7
972
FQ213847.1
-
7
973
FQ214087.1
-
7
973
FQ214354.1
-
7
972
FQ214553.1
-
7
974
FQ214850.1
-
7
972
FQ214997.1
-
7
972
FQ215007.1
-
7
972
FQ215662.1
-
7
972
FQ215682.1
-
7
972
FQ215870.1
-
7
972
FQ216120.1
-
7
973
FQ216296.1
-
7
972
FQ216389.1
-
7
973
FQ216406.1
-
7
976
FQ216495.1
-
7
972
FQ216618.1
-
7
972
FQ216655.1
-
7
973
FQ216733.1
-
7
974
FQ216774.1
-
7
972
FQ216813.1
-
7
971
FQ216851.1
-
7
972
FQ216913.1
-
7
973
FQ218561.1
-
7
972
FQ218832.1
-
7
972
FQ218943.1
-
7
972
FQ219025.1
-
7
975
FQ219184.1
-
7
976
FQ219389.1
-
7
972
FQ219457.1
-
7
972
FQ219678.1
-
7
877
FQ219815.1
-
7
972
FQ219817.1
-
7
972
FQ219837.1
-
7
972
FQ219855.1
-
7
972
FQ219923.1
-
7
972
FQ220008.1
-
7
972
FQ220051.1
-
7
972
FQ220077.1
-
7
972
FQ220115.1
-
7
972
FQ220201.1
-
7
970
FQ220213.1
-
7
972
FQ220247.1
-
7
973
FQ220282.1
-
7
974
FQ220284.1
-
7
973
FQ220302.1
-
7
972
FQ220338.1
-
7
972
FQ220345.1
-
7
973
FQ220370.1
-
7
972
FQ220406.1
-
7
973
FQ220413.1
-
7
972
FQ220420.1
-
7
974
FQ220433.1
-
7
970
FQ220444.1
-
7
972
FQ220445.1
-
7
968
FQ220449.1
-
7
972
FQ220500.1
-
7
972
FQ220514.1
-
7
972
FQ220527.1
-
7
973
FQ220533.1
-
7
1004
FQ220542.1
-
7
973
FQ220545.1
-
7
972
FQ220634.1
-
7
972
FQ220654.1
-
7
972
FQ220663.1
-
7
972
FQ220684.1
-
7
973
FQ220769.1
-
7
972
FQ220787.1
-
7
972
FQ220801.1
-
7
972
FQ220833.1
-
7
972
FQ220838.1
-
7
972
FQ220888.1
-
7
972
FQ220932.1
-
7
973
FQ221146.1
-
7
972
FQ221380.1
-
7
972
FQ221533.1
-
7
973
FQ221556.1
-
7
970
FQ222171.1
-
7
972
FQ222181.1
-
7
972
FQ222327.1
-
7
972
FQ222772.1
-
7
972
FQ222906.1
-
7
972
FQ223121.1
-
7
972
FQ223487.1
-
7
972
FQ224834.1
-
7
972
FQ224846.1
-
7
972
FQ225049.1
-
7
972
FQ225170.1
-
7
972
FQ225608.1
-
7
972
FQ225654.1
-
7
974
FQ225860.1
-
7
974
FQ225886.1
-
7
972
FQ226306.1
-
7
974
FQ227381.1
-
7
973
FQ227522.1
-
7
967
FQ227719.1
-
7
972
FQ227837.1
-
7
972
FQ227937.1
-
7
973
FQ227941.1
-
7
972
FQ227942.1
-
7
1092
FQ228548.1
-
7
973
FQ228995.1
-
7
972
FQ228996.1
-
7
972
FQ229139.1
-
7
974
FQ229159.1
-
7
972
FQ229164.1
-
7
973
FQ229208.1
-
7
972
FQ229241.1
-
7
972
FQ229244.1
-
7
972
FQ229248.1
-
5
852
FQ229272.1
-
7
972
FQ229273.1
-
7
973
FQ229303.1
-
7
972
FQ229337.1
-
7
972
FQ229352.1
-
7
972
FQ229365.1
-
7
972
FQ229397.1
-
7
972
FQ229408.1
-
7
973
FQ229433.1
-
7
972
FQ229448.1
-
7
972
FQ229521.1
-
7
972
FQ229547.1
-
7
972
FQ229554.1
-
7
972
FQ229566.1
-
7
968
FQ229610.1
-
7
972
FQ229626.1
-
7
969
FQ229726.1
-
7
972
FQ229740.1
-
7
972
FQ229742.1
-
7
972
FQ229776.1
-
7
972
FQ229808.1
-
7
972
FQ229815.1
-
7
972
FQ229822.1
-
7
972
FQ229830.1
-
7
974
FQ229883.1
-
7
974
FQ229901.1
-
7
972
FQ229929.1
-
7
972
FQ229932.1
-
5
870
FQ229952.1
-
7
972
FQ229974.1
-
7
972
FQ230005.1
-
7
972
FQ230055.1
-
7
971
FQ230127.1
-
7
972
FQ230267.1
-
7
975
FQ230943.1
-
7
972
FQ231445.1
-
7
974
FQ231525.1
-
7
972
FQ231591.1
-
7
974
FQ231710.1
-
7
973
FQ231733.1
-
7
973
FQ231897.1
-
7
974
FQ231967.1
-
7
972
FQ232029.1
-
7
973
FQ232154.1
-
7
972
FQ232165.1
-
7
972
FQ232310.1
-
7
971
FQ232319.1
-
7
973
FQ232697.1
-
7
974
FQ232767.1
-
7
972
FQ232951.1
-
7
972
FQ233152.1
-
7
971
FQ233473.1
-
7
972
FQ233732.1
-
7
972
FQ233737.1
-
7
972
FQ233808.1
-
7
972
FQ234195.1
-
7
972
FQ234341.1
-
7
972
FQ234613.1
-
7
972
FQ234733.1
-
7
972
FQ234735.1
-
7
862
FQ234863.1
-
7
973
FQ235079.1
-
7
972
FQ235122.1
-
7
972
FQ235259.1
-
7
972
X15096.1
-
7
969
Z29530.1
-
6
928
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational elongation
ribosome biogenesis
cellular response to interleukin-4
structural constituent of ribosome
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0