Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SREBF1 SIRNA Assays ›
          Gene Symbol:
          SREBF1
          Gene Name:
          sterol regulatory element binding transcription factor 1
          Gene Aliases:
          SREBP-1c, SREBP1, SREBP1a, bHLHd1
          Chromosome Location:
          Chr.17: 17811349 - 17837017 on Build GRCh38
          UniGene ID:
          Hs.592123
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5231

          Purification
          Size
          Availability Inventoried
          Catalog # AM51331
          Price 6,301.00
          Your Price
          Online offer:
          6301.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SREBF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001005291.2 NP_001005291.1 20 4010
          NM_001321096.2 NP_001308025.1 19 3782
          NM_004176.4 NP_004167.3 19 3920
          XM_005256772.4 XP_005256829.1 19 3920
          XM_017024970.1 XP_016880459.1 17 3351
          XM_017024971.1 XP_016880460.1 15 2702
          GenBank mRNA AB209609.1 - 18 4386
          AK091131.1 - 19 3749
          AK095325.1 - 14 2759
          AK128320.1 - 17 3429
          AK293795.1 - 19 3620
          AK297113.1 - 17 3334
          BC023621.2 - 13 2344
          BC026962.1 - 13 2536
          BC057388.1 - 19 3911 5786483
          BC063281.1 - 20 3985 5264244
          U00968.1 - 19 3892

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of heart rate by chemical signal
          regulation of transcription from RNA polymerase II promoter
          lipid metabolic process
          aging
          circadian rhythm
          cholesterol metabolic process
          insulin receptor signaling pathway
          lipid biosynthetic process
          cellular response to starvation
          response to glucose
          positive regulation of triglyceride biosynthetic process
          regulation of fatty acid metabolic process
          lung development
          positive regulation of histone deacetylation
          regulation of protein stability
          response to food
          response to retinoic acid
          response to progesterone
          response to glucagon
          response to drug
          mRNA transcription from RNA polymerase II promoter
          fat cell differentiation
          positive regulation of cholesterol biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          response to cAMP
          cellular response to fatty acid
          regulation of mitophagy
          regulation of protein targeting to mitochondrion
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein kinase binding
          protein complex binding
          sterol response element binding
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0