Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CHP1 SIRNA Assays ›
Gene Symbol:
Chp1
Gene Name:
calcineurin-like EF-hand protein 1
Gene Aliases:
Chp, Wrch2
Chromosome Location:
Chr.3: 111354506 - 111389998 on Build Rnor_6.0
UniGene ID:
Rn.11041
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
52755
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Chp1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_024139.2
NP_077053.1
4
425
GenBank mRNA
AB070350.1
-
4
410
BC062029.1
-
4
425
5623922
U39875.1
-
4
671
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_019769, AB025217, AK169146, AK156588, BC064784, and others.
Gene Ontology Categories:
Function(s)
Process(es)
microtubule bundle formation
negative regulation of protein phosphorylation
negative regulation of protein kinase activity
protein export from nucleus
negative regulation of phosphatase activity
calcium ion regulated exocytosis
calcium-mediated signaling
membrane docking
cytoplasmic microtubule organization
negative regulation of protein ubiquitination
negative regulation of protein autophosphorylation
negative regulation of NF-kappaB transcription factor activity
positive regulation of sodium:proton antiporter activity
negative regulation of protein import into nucleus
transcytosis
protein stabilization
positive regulation of protein transport
protein oligomerization
regulation of intracellular pH
positive regulation of protein glycosylation
membrane organization
membrane fusion
negative regulation of calcineurin-NFAT signaling cascade
cellular response to acidic pH
positive regulation of protein targeting to membrane
regulation of neuron death
protein kinase inhibitor activity
transporter activity
calcium ion binding
protein binding
microtubule binding
kinase binding
calcium-dependent protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0