Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCL2 SIRNA Assays ›
          Gene Symbol:
          Ccl2
          Gene Name:
          chemokine (C-C motif) ligand 2
          Gene Aliases:
          MCP-1, Scya2, Sigje
          Chromosome Location:
          Chr.10: 69412065 - 69413863 on Build Rnor_6.0
          UniGene ID:
          Rn.4772
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 53623

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 283.00
          Your Price
          Online offer: 274.65
          274.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ccl2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031530.1 NP_113718.1 2 256
          GenBank mRNA AF058786.1 - 2 256
          M57441.1 - 2 220

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_011333, AK153520, AF065930, AK132590, BC145869, and others.

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          response to hypoxia
          positive regulation of leukocyte mediated cytotoxicity
          positive regulation of endothelial cell proliferation
          leukocyte migration involved in inflammatory response
          monocyte chemotaxis
          positive regulation of leukocyte migration
          positive regulation of cellular extravasation
          cellular calcium ion homeostasis
          inflammatory response
          cytoskeleton organization
          transforming growth factor beta receptor signaling pathway
          aging
          glial cell migration
          regulation of cell shape
          response to heat
          response to wounding
          response to mechanical stimulus
          response to bacterium
          response to gamma radiation
          regulation of vascular endothelial growth factor production
          positive regulation of macrophage chemotaxis
          response to activity
          negative regulation of angiogenesis
          cytokine-mediated signaling pathway
          positive regulation of cell-cell adhesion
          neutrophil chemotaxis
          organ regeneration
          lipopolysaccharide-mediated signaling pathway
          response to lipopolysaccharide
          response to progesterone
          positive regulation of tumor necrosis factor production
          cellular response to insulin stimulus
          positive regulation of collagen biosynthetic process
          response to vitamin B3
          negative regulation of glial cell apoptotic process
          helper T cell extravasation
          cellular response to drug
          cellular response to macrophage colony-stimulating factor stimulus
          cellular response to platelet-derived growth factor stimulus
          chemokinesis
          response to drug
          response to amino acid
          protein kinase B signaling
          negative regulation of neuron apoptotic process
          astrocyte cell migration
          cellular response to fibroblast growth factor stimulus
          response to ethanol
          positive regulation of cell adhesion
          response to antibiotic
          vascular endothelial growth factor receptor signaling pathway
          macrophage chemotaxis
          lymphocyte chemotaxis
          positive regulation of synaptic transmission
          positive regulation of T cell activation
          response to glucocorticoid
          positive regulation of nitric-oxide synthase biosynthetic process
          maternal process involved in female pregnancy
          maternal process involved in parturition
          chemokine-mediated signaling pathway
          cellular response to lipopolysaccharide
          cellular response to retinoic acid
          cellular response to ATP
          cellular response to interferon-gamma
          cellular response to interleukin-1
          cellular response to interleukin-6
          cellular response to tumor necrosis factor
          cellular response to lipoprotein particle stimulus
          cellular response to high density lipoprotein particle stimulus
          cellular response to organic cyclic compound
          cellular response to dexamethasone stimulus
          positive regulation of monocyte chemotaxis
          positive regulation of immune complex clearance by monocytes and macrophages
          positive regulation of calcium ion import
          positive regulation of protein targeting to membrane
          positive regulation of apoptotic cell clearance
          negative regulation of natural killer cell chemotaxis
          G-protein coupled receptor binding
          chemokine activity
          heparin binding
          CCR2 chemokine receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline