Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL1B SIRNA Assays ›
Gene Symbol:
Il1b
Gene Name:
interleukin 1 beta
Gene Aliases:
-
Chromosome Location:
Chr.3: 121876256 - 121882637 on Build Rnor_6.0
UniGene ID:
Rn.9869
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
53796
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il1b
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_031512.2
NP_113700.2
4
250
GenBank mRNA
BC091141.1
-
4
250
7365029
M98820.1
-
4
270
Back To Top
More Information
Panther Classification:
Molecular Function -
interleukin superfamily
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
MAPK cascade
activation of MAPK activity
fever generation
response to hypoxia
positive regulation of protein phosphorylation
chronic inflammatory response to antigenic stimulus
purine nucleobase metabolic process
inflammatory response
immune response
positive regulation of cytosolic calcium ion concentration
aging
response to nutrient
learning or memory
memory
estrogen metabolic process
negative regulation of cell proliferation
glycoprotein metabolic process
response to heat
response to carbohydrate
response to ozone
response to organonitrogen compound
response to gamma radiation
positive regulation of vascular endothelial growth factor production
positive regulation of gene expression
negative regulation of gene expression
negative regulation of glucose transport
positive regulation of cell death
negative regulation of glutamate secretion
response to organic cyclic compound
cytokine-mediated signaling pathway
pentacyclic triterpenoid metabolic process
hyaluronan biosynthetic process
neutrophil chemotaxis
polyketide metabolic process
ovulation
sequestering of triglyceride
positive regulation of fever generation
lipopolysaccharide-mediated signaling pathway
positive regulation of prostaglandin secretion
response to estradiol
response to lipopolysaccharide
interleukin-1 beta production
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-6 production
positive regulation of interleukin-8 production
positive regulation of stress-activated MAPK cascade
positive regulation of immature T cell proliferation in thymus
response to ATP
response to vitamin D
response to L-ascorbic acid
positive regulation of heterotypic cell-cell adhesion
social behavior
ectopic germ cell programmed cell death
positive regulation of myosin light chain kinase activity
response to stilbenoid
cellular response to drug
response to diuretic
response to statin
wound healing
positive regulation of T cell proliferation
positive regulation of NF-kappaB import into nucleus
response to drug
positive regulation of apoptotic process
regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of I-kappaB kinase/NF-kappaB signaling
response to morphine
negative regulation of MAP kinase activity
response to peptide hormone
protein kinase B signaling
positive regulation of JUN kinase activity
positive regulation of chemokine biosynthetic process
positive regulation of interleukin-2 biosynthetic process
positive regulation of interleukin-6 biosynthetic process
positive regulation of nitric oxide biosynthetic process
response to ethanol
negative regulation of neuron differentiation
positive regulation of glial cell differentiation
positive regulation of angiogenesis
negative regulation of lipid metabolic process
positive regulation of mitotic nuclear division
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
negative regulation of insulin receptor signaling pathway
positive regulation of astrocyte differentiation
positive regulation of phagocytosis
negative regulation of neurogenesis
regulation of insulin secretion
leukocyte migration
negative regulation of lipid catabolic process
positive regulation of lipid catabolic process
positive regulation of membrane protein ectodomain proteolysis
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
positive regulation of cell division
positive regulation of calcidiol 1-monooxygenase activity
negative regulation of adiponectin secretion
positive regulation of ERK1 and ERK2 cascade
monocyte aggregation
cellular response to lipopolysaccharide
cellular response to antibiotic
cellular response to mechanical stimulus
cellular response to organic substance
cellular response to glucose stimulus
cellular response to lipid
cellular response to fatty acid
cellular response to organic cyclic compound
cellular response to methotrexate
response to dexamethasone
positive regulation of monocyte chemotactic protein-1 production
positive regulation of neutrophil chemotaxis
extrinsic apoptotic signaling pathway in absence of ligand
negative regulation of branching morphogenesis of a nerve
negative regulation of neural precursor cell proliferation
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
cytokine activity
interleukin-1 receptor binding
protein domain specific binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline