Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLAGL1 SIRNA Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
LOT1, ZAC, ZAC1
Chromosome Location:
Chr.6: 143940300 - 144064599 on Build GRCh38
UniGene ID:
Hs.743225
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
5381
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLAGL1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001080951.2
NP_001074420.1
8
1054
NM_001080952.2
NP_001074421.1
7
1206
NM_001080953.2
NP_001074422.1
7
1160
NM_001080954.2
NP_001074423.1
7
1159
NM_001080955.2
NP_001074424.1
6
503
NM_001080956.2
NP_001074425.1
6
698
NM_001289037.1
NP_001275966.1
5
462
NM_001289038.1
NP_001275967.1
4
416
NM_001289039.1
NP_001275968.1
5
519
NM_001289040.1
NP_001275969.1
6
488
NM_001289041.1
NP_001275970.1
6
698
NM_001289042.1
NP_001275971.1
5
850
NM_001289043.1
NP_001275972.1
7
973
NM_001289044.1
NP_001275973.1
6
924
NM_001289045.1
NP_001275974.1
7
1468
NM_001289046.1
NP_001275975.1
9
1449
NM_001289047.1
NP_001275976.1
4
1013
NM_001289048.1
NP_001275977.1
7
1174
NM_001289049.1
NP_001275978.1
6
1119
NM_001317156.1
NP_001304085.1
4
1380
NM_001317157.1
NP_001304086.1
3
1556
NM_001317158.1
NP_001304087.1
3
918
NM_001317159.1
NP_001304088.1
8
1040
NM_001317160.1
NP_001304089.1
6
684
NM_001317161.1
NP_001304090.1
7
1174
NM_001317162.1
NP_001304091.1
8
1200
NM_006718.4
NP_006709.2
7
1192
GenBank mRNA
AJ006354.1
-
3
1586
AJ303119.1
-
2
226
AJ311395.1
-
9
1439
AK091707.1
-
4
1380
AK096331.1
-
7
1436
AK314570.1
-
6
545
AK315184.1
-
4
714
BC074814.2
-
2
339
30915533
BC109085.2
-
3
646
40034491
BC109086.2
-
3
646
40034497
BX537397.1
-
3
3887
DA426770.1
-
6
545
DA675185.1
-
6
497
U72621.4
-
6
2531
U81992.1
-
3
310
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0