Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NRG1 SIRNA Assays ›
          Gene Symbol:
          Nrg1
          Gene Name:
          neuregulin 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.16: 62969988 - 64057434 on Build Rnor_6.0
          UniGene ID:
          Rn.37438
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 53846

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nrg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001271118.1 NP_001258047.1 2 1352
          NM_001271119.1 NP_001258048.1 2 1352
          NM_001271120.1 NP_001258049.1 2 1352
          NM_001271121.1 NP_001258050.1 4 805
          NM_001271122.1 NP_001258051.1 4 805
          NM_001271123.1 NP_001258052.1 6 907
          NM_001271124.1 NP_001258053.1 6 907
          NM_001271125.1 NP_001258054.1 6 907
          NM_001271126.1 NP_001258055.1 6 907
          NM_001271127.1 NP_001258056.1 6 907
          NM_001271128.1 NP_001258057.1 6 907
          NM_001271129.1 NP_001258058.1 6 907
          NM_001271130.1 NP_001258059.1 6 977
          NM_031588.2 NP_113776.2 6 907
          GenBank mRNA AF194438.1 - 2 1366
          AF194439.1 - 2 1100
          AF194440.1 - 2 175
          AF194441.1 - 2 191
          AF194442.1 - 2 191
          AF194443.1 - 2 609
          AF194993.1 - 6 977
          AF194995.1 - 6 810
          AF194996.1 - 6 810
          AF194997.1 - 6 810
          AY973241.1 - 4 299
          AY973243.1 - 4 299
          AY973244.1 - 4 263
          AY973245.1 - 6 573
          AY995221.1 - 6 563
          AY995222.1 - 4 461
          AY995223.1 - 4 461
          DQ176766.1 - 2 731
          M92430.1 - 6 607
          U02315.1 - 6 532
          U02316.1 - 6 907
          U02317.1 - 6 907
          U02318.1 - 6 907
          U02319.1 - 6 907
          U02320.1 - 6 907
          U02321.1 - 6 907
          U02322.1 - 6 907
          U02323.1 - 6 907
          U02324.1 - 6 607

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_013960, NM_001160007, NM_001159995, NM_001160004, NM_004495, and others.
          This siRNA also targets the following transcripts in Mouse: NM_178591, AY227027, AK141037, BC151113, AK048708, and others.

          Panther Classification:

          Molecular Function -

          growth factor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          cell morphogenesis
          cardiac conduction system development
          ventricular trabecula myocardium morphogenesis
          transmembrane receptor protein tyrosine kinase signaling pathway
          activation of transmembrane receptor protein tyrosine kinase activity
          nervous system development
          synapse assembly
          peripheral nervous system development
          heart development
          muscle organ development
          locomotory behavior
          regulation of blood pressure
          cell proliferation
          glial cell differentiation
          negative regulation of cardiac muscle cell apoptotic process
          glucose transport
          cell migration
          glial cell fate commitment
          cell differentiation
          positive regulation of cell growth
          intracellular signal transduction
          ERBB signaling pathway
          wound healing
          negative regulation of protein catabolic process
          myelination
          regulation of protein heterodimerization activity
          cellular protein complex disassembly
          neurotransmitter receptor metabolic process
          regulation of cell differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of Ras protein signal transduction
          animal organ development
          neuron fate commitment
          negative regulation of secretion
          positive regulation of protein kinase B signaling
          ventricular cardiac muscle cell differentiation
          positive regulation of cardiac muscle cell proliferation
          cardiac muscle cell myoblast differentiation
          endocardial cell differentiation
          positive regulation of protein tyrosine kinase activity
          positive regulation of calcineurin-NFAT signaling cascade
          positive regulation of cardiac muscle cell differentiation
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          transcription cofactor activity
          receptor binding
          ErbB-2 class receptor binding
          growth factor activity
          protein tyrosine kinase activator activity
          ErbB-3 class receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline