Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS2 SIRNA Assays ›
          Gene Symbol:
          Rps2
          Gene Name:
          ribosomal protein S2
          Gene Aliases:
          Rps2r1
          Chromosome Location:
          Chr.10: 14088171 - 14090020 on Build Rnor_6.0
          UniGene ID:
          Rn.2115
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 54587
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_031838.3 NP_114026.3 3 331
          GenBank mRNA BC098726.1 - 3 320 7378399
          CB324420.1 - 4 350
          CD372915.1 - 3 331
          FQ209478.1 - 4 342
          FQ210378.1 - 4 343
          FQ213061.1 - 4 343
          FQ213116.1 - 4 340
          FQ214025.1 - 4 343
          FQ215811.1 - 4 345
          FQ216756.1 - 4 343
          FQ218550.1 - 4 344
          FQ218641.1 - 4 342
          FQ218643.1 - 4 346
          FQ219176.1 - 4 342
          FQ219789.1 - 4 343
          FQ219844.1 - 4 345
          FQ220315.1 - 4 343
          FQ220515.1 - 4 343
          FQ220642.1 - 4 343
          FQ220831.1 - 4 342
          FQ220841.1 - 4 343
          FQ220885.1 - 4 343
          FQ220902.1 - 4 343
          FQ221771.1 - 4 343
          FQ221977.1 - 4 342
          FQ222558.1 - 4 343
          FQ222607.1 - 4 342
          FQ222729.1 - 4 343
          FQ222900.1 - 4 343
          FQ223242.1 - 4 343
          FQ225100.1 - 5 216
          FQ225937.1 - 4 340
          FQ226432.1 - 4 345
          FQ226962.1 - 4 343
          FQ228202.1 - 4 343
          FQ229347.1 - 4 343
          FQ229368.1 - 4 343
          FQ229383.1 - 4 341
          FQ229400.1 - 4 342
          FQ229773.1 - 3 340
          FQ229778.1 - 4 343
          FQ230085.1 - 4 342
          FQ230109.1 - 4 345
          FQ230137.1 - 4 343
          FQ230697.1 - 4 340
          FQ231866.1 - 4 343
          FQ233653.1 - 4 343
          FQ234367.1 - 4 343
          FQ234813.1 - 4 343
          FQ235141.1 - 4 345
          U92696.1 - 4 224
          U92697.1 - 4 247
          U92698.1 - 4 250
          U92699.1 - 4 223
          U92700.1 - 4 196
          X57432.1 - 4 341

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal small subunit assembly
          formation of translation preinitiation complex
          positive regulation of transferase activity
          cellular response to interleukin-4
          RNA binding
          mRNA binding
          structural constituent of ribosome
          fibroblast growth factor binding
          enzyme binding
          protein complex binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-254dz:80/100.66.77.50:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline