Search
Search

Fmr1| Gene Symbol: Fmr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052804.1 | NP_434691.1 | 4, 5 | 453 | ||
| XM_006229530.2 | XP_006229592.1 | 4, 5 | 503 | |||
| XM_006229531.2 | XP_006229593.1 | 4, 5 | 503 | |||
| XM_006229532.2 | XP_006229594.1 | 4, 5 | 503 | |||
| XM_006229535.2 | XP_006229597.1 | 3, 4 | 222 | |||
| XM_008773597.1 | XP_008771819.1 | 4, 5 | 335 | |||
| GenBank mRNA | AF435434.1 | - | 4, 5 | 272 | ||
| AY240947.1 | - | 4, 5 | 453 | |||
| U60145.1 | - | 2, 3 | 162 | |||