Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS27 SIRNA Assays ›
          Gene Symbol:
          Rps27 LOC100359517 LOC100362987 LOC100911728
          Gene Name:
          ribosomal protein S27
          ribosomal protein S27-like
          ribosomal protein S27-like
          40S ribosomal protein S27-like
          Gene Aliases:
          -
          -
          -
          -
          Chromosome Location:
          Chr.2: 189572175 - 189573280 on Build Rnor_6.0
          Chromosome Location:
          Chr.14: 104396613 - 104451972 on Build Rnor_6.0
          Chromosome Location:
          Chr.9: 10737527 - 10737830 on Build Rnor_6.0
          Chromosome Location:
          Chr.13: 89218835 - 89219225 on Build Rnor_6.0
          UniGene ID:
          Rn.144623
          -
          -
          -
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID 55431

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053597.3 NP_446049.1 3, 4 256
          GenBank mRNA AF184893.1 - 3, 4 250
          BC061539.1 - 260 6918361
          FQ209846.1 - 3, 4 256
          FQ210151.1 - 3, 4 255
          FQ217607.1 - 3, 4 258
          FQ217624.1 - 3, 4 256
          FQ218228.1 - 3, 4 257
          FQ218581.1 - 3, 4 256
          FQ221131.1 - 3, 4 256
          FQ221294.1 - 3, 4 256
          FQ221565.1 - 3, 4 256
          FQ221645.1 - 3, 4 255
          FQ221664.1 - 3, 4 256
          FQ221723.1 - 3, 4 255
          FQ221727.1 - 3, 4 253
          FQ221743.1 - 3, 4 256
          FQ221809.1 - 3, 4 256
          FQ221911.1 - 3, 4 256
          FQ221975.1 - 3, 4 257
          FQ222049.1 - 3, 4 256
          FQ222054.1 - 3, 4 257
          FQ222270.1 - 3, 4 257
          FQ222348.1 - 4, 5 373
          FQ222848.1 - 3, 4 256
          FQ222865.1 - 3, 4 256
          FQ222911.1 - 3, 4 256
          FQ223072.1 - 3, 4 256
          FQ223291.1 - 3, 4 255
          FQ223549.1 - 3, 4 256
          FQ223862.1 - 3, 4 256
          FQ224473.1 - 3, 4 256
          FQ228507.1 - 3, 4 257
          FQ228599.1 - 3, 4 256
          FQ228823.1 - 3, 4 256
          FQ229526.1 - 3, 4 256
          Gene Symbol: LOC100359517
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_008770451.1 XP_008768673.1 2 304
          Gene Symbol: LOC100362987
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_002730094.4 XP_002730140.1 1 223
          Gene Symbol: LOC100911728
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_003751271.3 XP_003751319.1 1 305

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: NM_027015, NM_001190258, BC048352, BC055693, AK166551, and others.

          Panther Classification:

          Molecular Function -

          ribosomal protein translational protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          biological_process
          structural constituent of ribosome
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0