Search
Search

Acsl4| Gene Symbol: Acsl4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053623.1 | NP_446075.1 | 15 | 1970 | ||
| XM_006257314.2 | XP_006257376.1 | 15 | 2308 | |||
| XM_006257315.2 | XP_006257377.1 | 16 | 2089 | |||
| XM_006257316.2 | XP_006257378.1 | 15 | 2113 | |||
| XM_008773385.1 | XP_008771607.1 | 16 | 2382 | |||
| GenBank mRNA | D85189.1 | - | 15 | 1970 | ||