Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ST18 SIRNA Assays ›
          Gene Symbol:
          ST18
          Gene Name:
          ST18, C2H2C-type zinc finger
          Gene Aliases:
          NZF3, ZC2H2C3, ZC2HC10, ZNF387
          Chromosome Location:
          Chr.8: 52110832 - 52409879 on Build GRCh38
          UniGene ID:
          Hs.655499
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5606

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ST18
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014682.2 NP_055497.1 8, 9 906
          XM_006716487.1 XP_006716550.1 8, 9 673
          XM_011517629.1 XP_011515931.1 9, 10 829
          XM_011517631.1 XP_011515933.1 10, 11 870
          XM_011517632.1 XP_011515934.1 8, 9 684
          XM_011517633.1 XP_011515935.1 8, 9 677
          XM_011517634.1 XP_011515936.1 8, 9 873
          XM_011517635.1 XP_011515937.1 8, 9 877
          XM_011517636.2 XP_011515938.1 7, 8 954
          XM_011517637.1 XP_011515939.1 6, 7 411
          XM_011517638.2 XP_011515940.1 5, 6 452
          XM_011517642.1 XP_011515944.1 9, 10 829
          XM_017014047.1 XP_016869536.1 8, 9 1033
          XM_017014048.1 XP_016869537.1 8, 9 1037
          XM_017014049.1 XP_016869538.1 9, 10 1083
          XM_017014050.1 XP_016869539.1 8, 9 870
          XM_017014051.1 XP_016869540.1 7, 8 877
          XM_017014052.1 XP_016869541.1 9, 10 1087
          XM_017014053.1 XP_016869542.1 9, 10 1074
          XM_017014054.1 XP_016869543.1 7, 8 1123
          XM_017014055.1 XP_016869544.1 7, 8 714
          XM_017014056.1 XP_016869545.1 10, 11 1124
          XM_017014057.1 XP_016869546.1 8, 9 927
          XM_017014058.1 XP_016869547.1 8, 9 688
          XM_017014059.1 XP_016869548.1 8, 9 874
          XM_017014060.1 XP_016869549.1 9, 10 911
          XM_017014061.1 XP_016869550.1 7, 8 1046
          XM_017014062.1 XP_016869551.1 6, 7 798
          XM_017014063.1 XP_016869552.1 7, 8 1344
          XM_017014064.1 XP_016869553.1 7, 8 958
          XM_017014065.1 XP_016869554.1 9, 10 1078
          XM_017014066.1 XP_016869555.1 7, 8 717
          XM_017014067.1 XP_016869556.1 7, 8 1127
          XM_017014068.1 XP_016869557.1 8, 9 918
          XM_017014069.1 XP_016869558.1 5, 6 615
          XM_017014070.1 XP_016869559.1 4, 5 536
          XM_017014071.1 XP_016869560.1 5, 6 784
          XM_017014072.1 XP_016869561.1 8, 9 1035
          XM_017014073.1 XP_016869562.1 8, 9 673
          XM_017014074.1 XP_016869563.1 7, 8 956
          XM_017014081.1 XP_016869570.1 8, 9 1040
          XM_017014082.1 XP_016869571.1 8, 9 875
          XM_017014083.1 XP_016869572.1 7, 8 715
          XM_017014084.1 XP_016869573.1 5, 6 453
          GenBank mRNA AB011107.1 - 8, 9 770
          BC031075.1 - 8, 9 803
          BC117147.1 - 3, 4 163 40125698
          BC117149.1 - 3, 4 163 40125700
          BC143872.1 - 3, 4 162

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          zinc ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-kjp6x:80/100.66.78.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0