Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPL6 SIRNA Assays ›
          Gene Symbol:
          Rpl6
          Gene Name:
          ribosomal protein L6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.12: 40877578 - 40882030 on Build Rnor_6.0
          UniGene ID:
          Rn.234072
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 56226

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rpl6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_053971.3 NP_446423.2 5 655
          XM_006249377.1 XP_006249439.1 6 674
          XM_006249378.2 XP_006249440.1 6 708
          GenBank mRNA BC061784.1 - 5 651
          BC078761.1 - 5 658 7123086
          BC097295.1 - 5 659 7446680
          FQ213055.1 - 4 333
          FQ213078.1 - 4 333
          FQ216758.1 - 4 333
          FQ217744.1 - 4 333
          FQ219956.1 - 4 333
          FQ219982.1 - 4 333
          FQ220180.1 - 4 333
          FQ220346.1 - 4 333
          FQ220362.1 - 4 333
          FQ220471.1 - 4 333
          FQ220678.1 - 4 333
          FQ220699.1 - 4 333
          FQ220887.1 - 4 333
          FQ220959.1 - 4 333
          FQ221615.1 - 4 333
          FQ222080.1 - 4 333
          FQ222331.1 - 4 333
          FQ222465.1 - 4 333
          FQ222658.1 - 4 333
          FQ223301.1 - 4 333
          FQ226632.1 - 4 334
          FQ227060.1 - 4 333
          FQ227100.1 - 4 333
          FQ227955.1 - 4 333
          FQ227974.1 - 4 333
          FQ228583.1 - 4 333
          FQ228665.1 - 4 333
          FQ229345.1 - 4 333
          FQ229355.1 - 4 333
          FQ229484.1 - 6 673
          FQ229571.1 - 4 333
          FQ229624.1 - 4 333
          FQ229675.1 - 4 333
          FQ229700.1 - 4 333
          FQ229820.1 - 4 333
          FQ229854.1 - 4 333
          FQ229857.1 - 4 333
          FQ229943.1 - 6 672
          FQ230171.1 - 4 333
          FQ230222.1 - 4 333
          FQ233198.1 - 4 333
          FQ233475.1 - 4 333
          FQ234880.1 - 6 672
          FQ235095.1 - 4 333
          X87107.1 - 6 669

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ribosomal protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          structural constituent of ribosome
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0