Search
Search

Rab7l1| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133590.1 | NP_598274.1 | 3 | 663 | ||
| XM_006249744.2 | XP_006249806.1 | 4 | 640 | |||
| XM_006249745.2 | XP_006249807.1 | 4 | 497 | |||
| XM_008769422.1 | XP_008767644.1 | 2 | 193 | |||
| GenBank mRNA | BC081732.1 | - | 4 | 619 | 7109012 | |
| FQ233577.1 | - | 3 | 429 | |||
| FQ234855.1 | - | 3 | 429 | |||
| X96663.1 | - | 3 | 663 | |||