Search
Search

Rab7l1| Gene Symbol: Rab7l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133590.1 | NP_598274.1 | 5 | 913 | ||
| XM_006249744.2 | XP_006249806.1 | 6 | 890 | |||
| XM_006249745.2 | XP_006249807.1 | 6 | 747 | |||
| XM_008769422.1 | XP_008767644.1 | 4 | 443 | |||
| GenBank mRNA | BC081732.1 | - | 5 | 1156 | 7109012 | |
| FQ233577.1 | - | 5 | 679 | |||
| FQ234855.1 | - | 5 | 679 | |||
| X96663.1 | - | 5 | 913 | |||