Search
Search

Camk1| Gene Symbol: Camk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134468.1 | NP_604463.1 | 3 | 239 | ||
| XM_006236999.2 | XP_006237061.1 | 4 | 908 | |||
| XM_006237000.2 | XP_006237062.1 | 4 | 904 | |||
| XM_006237001.2 | XP_006237063.1 | 3 | 771 | |||
| XM_006237002.2 | XP_006237064.1 | 3 | 698 | |||
| XM_006237003.2 | XP_006237065.1 | 3 | 697 | |||
| GenBank mRNA | BC071177.1 | - | 3 | 174 | 7098592 | |
| FQ213485.1 | - | 202 | ||||
| L24907.1 | - | 3 | 213 | |||
| L26288.1 | - | 3 | 239 | |||