Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IFNG SIRNA Assays ›
Gene Symbol:
Ifng
Gene Name:
interferon gamma
Gene Aliases:
IFNG2
Chromosome Location:
Chr.7: 61337383 - 61341419 on Build Rnor_6.0
UniGene ID:
Rn.10795
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
57800
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ifng
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_138880.2
NP_620235.1
2, 3
197
GenBank mRNA
AF010466.1
-
2, 3
197
Back To Top
More Information
Panther Classification:
Molecular Function -
cytokine
interferon superfamily
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
protein import into nucleus, translocation
negative regulation of transcription from RNA polymerase II promoter
neutrophil apoptotic process
regulation of the force of heart contraction
adaptive immune response
CD8-positive, alpha-beta T cell differentiation involved in immune response
regulation of transcription, DNA-templated
apoptotic process
inflammatory cell apoptotic process
humoral immune response
cell cycle arrest
positive regulation of cell proliferation
negative regulation of cell proliferation
response to virus
antigen processing and presentation
neutrophil chemotaxis
negative regulation of epithelial cell differentiation
endoplasmic reticulum unfolded protein response
negative regulation of myelination
positive regulation of synaptic transmission, cholinergic
negative regulation of interleukin-17 production
positive regulation of interleukin-12 production
positive regulation of interleukin-23 production
positive regulation of tumor necrosis factor production
positive regulation of peptidyl-serine phosphorylation of STAT protein
positive regulation of smooth muscle cell apoptotic process
positive regulation of T cell proliferation
response to drug
positive regulation of tyrosine phosphorylation of Stat1 protein
defense response to bacterium
defense response to protozoan
negative regulation of growth of symbiont in host
negative regulation of growth of symbiont involved in interaction with host
positive regulation of chemokine biosynthetic process
positive regulation of interleukin-12 biosynthetic process
positive regulation of MHC class II biosynthetic process
positive regulation of interleukin-6 biosynthetic process
positive regulation of nitric oxide biosynthetic process
positive regulation of neuron differentiation
positive regulation of osteoclast differentiation
positive regulation of cell adhesion
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of isotype switching to IgG isotypes
negative regulation of smooth muscle cell proliferation
positive regulation of interleukin-1 beta secretion
regulation of immune response
regulation of insulin secretion
sensory perception of mechanical stimulus
positive regulation of membrane protein ectodomain proteolysis
defense response to virus
positive regulation of killing of cells of other organism
positive regulation of fructose 1,6-bisphosphate 1-phosphatase activity
positive regulation of fructose 1,6-bisphosphate metabolic process
positive regulation of vitamin D biosynthetic process
positive regulation of calcidiol 1-monooxygenase activity
cellular response to lipopolysaccharide
cellular response to interleukin-18
extrinsic apoptotic signaling pathway
regulation of neuronal action potential
positive regulation of tumor necrosis factor (ligand) superfamily member 11 production
regulation of hepatocyte proliferation
cytokine activity
interferon-gamma receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0