Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ADRB1 SIRNA Assays ›
Gene Symbol:
Adrb1
Gene Name:
adrenoceptor beta 1
Gene Aliases:
B1AR, RATB1AR
Chromosome Location:
Chr.1: 277537585 - 277538985 on Build Rnor_6.0
UniGene ID:
Rn.87064
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
59243
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Adrb1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012701.1
NP_036833.1
1
764
Back To Top
More Information
Set Membership:
G-protein coupled receptor
Panther Classification:
Molecular Function -
G-protein coupled receptor
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of heart rate by epinephrine-norepinephrine
positive regulation of the force of heart contraction by epinephrine-norepinephrine
diet induced thermogenesis
vasodilation by norepinephrine-epinephrine involved in regulation of systemic arterial blood pressure
positive regulation of the force of heart contraction by norepinephrine
positive regulation of systemic arterial blood pressure
glycogen catabolic process
apoptotic process
G-protein coupled receptor signaling pathway
adenylate cyclase-activating G-protein coupled receptor signaling pathway
Rho protein signal transduction
aging
memory
response to cold
positive regulation of heart rate
sensory perception of pain
positive regulation of cAMP biosynthetic process
heat generation
protein localization to organelle
negative regulation of urine volume
negative regulation of multicellular organism growth
wound healing
fear response
positive regulation of apoptotic process
positive regulation of cAMP-mediated signaling
positive regulation of adenylate cyclase activity
negative regulation of smooth muscle contraction
positive regulation of saliva secretion
brown fat cell differentiation
regulation of calcium ion transport
lipid homeostasis
inhibitory postsynaptic potential
positive regulation of cell growth involved in cardiac muscle cell development
adrenergic receptor signaling pathway
adenylate cyclase-activating adrenergic receptor signaling pathway
regulation of cardiac muscle cell contraction
positive regulation of renin secretion into blood stream
positive regulation of long-term synaptic potentiation
positive regulation of cation channel activity
beta1-adrenergic receptor activity
receptor signaling protein activity
Ras guanyl-nucleotide exchange factor activity
protein binding
drug binding
PDZ domain binding
alpha-2A adrenergic receptor binding
dopamine binding
protein heterodimerization activity
epinephrine binding
norepinephrine binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0