Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DEGS1 SIRNA Assays ›
          Gene Symbol:
          Degs1
          Gene Name:
          degenerative spermatocyte homolog 1 (Drosophila)
          Gene Aliases:
          AA536663, Degs, Des1, Mdes
          Chromosome Location:
          Chr.1: 182275770 - 182282759 on Build GRCm38
          UniGene ID:
          Mm.29648
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61148

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Degs1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007853.4 NP_031879.1 2 537
          GenBank mRNA AF466376.1 - 2 554
          AK002617.1 - 2 545
          AK077257.1 - 2 538
          AK149811.1 - 2 527
          AK150908.1 - 2 526
          AK163404.1 - 2 527
          AK165352.1 - 2 552
          BC003751.1 - 2 555 3600571
          Y08460.1 - 2 522

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydroxylase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          fatty acid biosynthetic process
          sphingolipid biosynthetic process
          positive regulation of apoptotic process
          ceramide biosynthetic process
          oxidation-reduction process
          oxidoreductase activity
          fluorene oxygenase activity
          mono-butyltin dioxygenase activity
          tri-n-butyltin dioxygenase activity
          di-n-butyltin dioxygenase activity
          methylsilanetriol hydroxylase activity
          methyl tertiary butyl ether 3-monooxygenase activity
          4-nitrocatechol 4-monooxygenase activity
          4-chlorophenoxyacetate monooxygenase activity
          tert-butanol 2-monooxygenase activity
          alpha-pinene monooxygenase activity
          dimethylsilanediol hydroxylase activity
          ammonia monooxygenase activity
          hydroxymethylsilanetriol oxidase activity
          2-hydroxyisobutyrate 3-monooxygenase activity
          alpha-pinene dehydrogenase activity
          bisphenol A hydroxylase B activity
          2,2-bis(4-hydroxyphenyl)-1-propanol hydroxylase activity
          9-fluorenone-3,4-dioxygenase activity
          anthracene 9,10-dioxygenase activity
          2-(methylthio)benzothiazole monooxygenase activity
          2-hydroxybenzothiazole monooxygenase activity
          benzothiazole monooxygenase activity
          2,6-dihydroxybenzothiazole monooxygenase activity
          pinacolone 5-monooxygenase activity
          thioacetamide S-oxygenase activity
          thioacetamide S-oxide S-oxygenase activity
          endosulfan monooxygenase I activity
          N-nitrodimethylamine hydroxylase activity
          4-(1-ethyl-1,4-dimethyl-pentyl)phenol monoxygenase activity
          endosulfan ether monooxygenase activity
          pyrene 4,5-monooxygenase activity
          pyrene 1,2-monooxygenase activity
          1-hydroxypyrene 6,7-monooxygenase activity
          1-hydroxypyrene 7,8-monooxygenase activity
          phenylboronic acid monooxygenase activity
          sphingolipid delta-4 desaturase activity
          spheroidene monooxygenase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0