Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DLG1 SIRNA Assays ›
          Gene Symbol:
          Dlg1
          Gene Name:
          discs, large homolog 1 (Drosophila)
          Gene Aliases:
          B130052P05Rik, Dlgh1, E-dlg/SAP97, SAP-97, SAP97, mKIAA4187
          Chromosome Location:
          Chr.16: 31663443 - 31873356 on Build GRCm38
          UniGene ID:
          Mm.382
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61243

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Dlg1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252433.1 NP_001239362.1 13 1789
          NM_001252434.1 NP_001239363.1 13 1789
          NM_001252435.1 NP_001239364.1 14 1547
          NM_001252436.1 NP_001239365.1 10 1654
          NM_007862.3 NP_031888.2 14 1888
          GenBank mRNA AK142893.1 - 10 1655
          AK143780.1 - 13 1576
          AK164228.1 - 5 557
          AK220189.1 - 13 1724
          AY159380.1 - 13 1449
          BC047142.1 - 14 1547 6396595
          BC057118.1 - 14 1784 6410247
          CF164686.1 - 5 564
          U93309.1 - 13 1478

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          ureteric bud development
          branching involved in ureteric bud morphogenesis
          immunological synapse formation
          endothelial cell proliferation
          lens development in camera-type eye
          T cell cytokine production
          actin filament organization
          chemical synaptic transmission
          nervous system development
          protein localization
          positive regulation of cell proliferation
          single organismal cell-cell adhesion
          peristalsis
          positive regulation of actin filament polymerization
          cortical actin cytoskeleton organization
          membrane raft organization
          regulation of myelination
          activation of protein kinase activity
          regulation of protein localization
          positive regulation of multicellular organism growth
          T cell activation
          negative regulation of T cell proliferation
          regulation of membrane potential
          amyloid precursor protein metabolic process
          receptor clustering
          positive regulation of potassium ion transport
          establishment or maintenance of epithelial cell apical/basal polarity
          negative regulation of mitotic cell cycle
          nucleotide phosphorylation
          reproductive structure development
          positive regulation of developmental growth
          embryonic skeletal system morphogenesis
          tissue morphogenesis
          smooth muscle tissue development
          negative regulation of epithelial cell proliferation
          negative regulation of protein kinase B signaling
          hard palate development
          bicellular tight junction assembly
          protein localization to plasma membrane
          positive regulation of establishment of protein localization to plasma membrane
          receptor localization to synapse
          guanylate kinase activity
          protein binding
          protein C-terminus binding
          potassium channel regulator activity
          kinase binding
          protein kinase binding
          phosphatase binding
          PDZ domain binding
          mitogen-activated protein kinase kinase binding
          protein complex scaffold
          ionotropic glutamate receptor binding
          ion channel binding
          L27 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0