Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EZH2 SIRNA Assays ›
          Gene Symbol:
          Ezh2
          Gene Name:
          enhancer of zeste homolog 2 (Drosophila)
          Gene Aliases:
          Enx-1, Enx1h, KMT6, mKIAA4065
          Chromosome Location:
          Chr.6: 47530274 - 47595340 on Build GRCm38
          UniGene ID:
          Mm.246688
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61343

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ezh2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146689.1 NP_001140161.1 2 244
          NM_007971.2 NP_031997.2 2 244
          GenBank mRNA AK088575.1 - 2 265
          AK153835.1 - 1 165
          AK157853.1 - 2 245
          AK220174.1 - 2 199
          BC003772.1 - 2 153 3492110
          BC016391.1 - 2 166 3586689
          BC079538.1 - 2 195 6817366
          U52951.1 - 2 234

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          histone modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          regulation of protein phosphorylation
          DNA methylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          regulation of gene expression
          negative regulation of gene expression
          positive regulation of epithelial to mesenchymal transition
          regulation of gliogenesis
          skeletal muscle satellite cell maintenance involved in skeletal muscle regeneration
          chromatin modification
          histone methylation
          peptidyl-lysine methylation
          cerebellar cortex development
          methylation
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of cell proliferation
          regulation of circadian rhythm
          positive regulation of MAP kinase activity
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of GTPase activity
          negative regulation of epidermal cell differentiation
          negative regulation of gene expression, epigenetic
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          rhythmic process
          regulation of neurogenesis
          negative regulation of striated muscle cell differentiation
          cellular response to hydrogen peroxide
          G1 to G0 transition
          histone H3-K27 methylation
          protein localization to chromatin
          positive regulation of protein serine/threonine kinase activity
          positive regulation of dendrite development
          negative regulation of G1/S transition of mitotic cell cycle
          regulatory region DNA binding
          core promoter binding
          chromatin binding
          RNA binding
          protein binding
          methyltransferase activity
          protein-lysine N-methyltransferase activity
          transferase activity
          histone-lysine N-methyltransferase activity
          chromatin DNA binding
          histone methyltransferase activity
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0