Search
Search

Fdx1| Gene Symbol: Fdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007996.2 | NP_032022.1 | 4 | 629 | ||
| GenBank mRNA | AK004879.1 | - | 4 | 583 | ||
| AK054524.1 | - | 1 | 2260 | |||
| BC099518.1 | - | 4 | 483 | |||
| BC132173.1 | - | 4 | 554 | 40130450 | ||
| BC132175.1 | - | 4 | 554 | 40130452 | ||
| D43689.1 | - | 4 | 560 | |||
| D43690.1 | - | 4 | 555 | |||
| L29123.1 | - | 4 | 582 | |||