Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGFR3 SIRNA Assays ›
          Gene Symbol:
          Fgfr3
          Gene Name:
          fibroblast growth factor receptor 3
          Gene Aliases:
          CD333, FR3, Fgfr-3, Flg-2, HBGFR, Mfr3, sam3
          Chromosome Location:
          Chr.5: 33721724 - 33737068 on Build GRCm38
          UniGene ID:
          Mm.6904
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61560

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgfr3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001163215.2 NP_001156687.1 14 2205
          NM_001163216.2 NP_001156688.1 12 1920
          NM_001163217.2 NP_001156689.1 13 1980
          NM_001205270.1 NP_001192199.1 14 2182
          NM_008010.5 NP_032036.2 14 2179
          GenBank mRNA AF024638.1 - 12 1828
          AK143592.1 - 14 2115
          AK163495.1 - 12 1758
          BC053056.1 - 14 2089 5708838
          M81342.1 - 14 2097
          S56291.1 - 13 1924
          X58255.1 - 14 2083

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: NM_022963, NM_213647, NM_002011, M59373, AK225909, and others.
          This siRNA also targets the following transcripts in Rat: NM_053429, XM_006251396, XM_006251395, XM_006251390, XM_006251393, and others.

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          MAPK cascade
          skeletal system development
          positive regulation of endothelial cell proliferation
          morphogenesis of an epithelium
          lens morphogenesis in camera-type eye
          protein phosphorylation
          apoptotic process
          cell-cell signaling
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          positive regulation of phospholipase activity
          oligodendrocyte development
          phosphorylation
          peptidyl-tyrosine phosphorylation
          substantia nigra development
          central nervous system myelination
          cell differentiation
          bone mineralization
          forebrain development
          positive regulation of protein ubiquitination
          somatic stem cell population maintenance
          post-anal tail morphogenesis
          positive regulation of tyrosine phosphorylation of Stat1 protein
          positive regulation of tyrosine phosphorylation of Stat3 protein
          positive regulation of MAPK cascade
          positive regulation of neuron apoptotic process
          positive regulation of phosphatidylinositol 3-kinase activity
          positive regulation of cell differentiation
          negative regulation of mitotic nuclear division
          negative regulation of smoothened signaling pathway
          protein autophosphorylation
          digestive tract morphogenesis
          negative regulation of developmental growth
          response to axon injury
          negative regulation of astrocyte differentiation
          inner ear development
          negative regulation of epithelial cell proliferation
          positive regulation of peptidyl-tyrosine phosphorylation
          cartilage development
          inner ear receptor cell differentiation
          bone morphogenesis
          axonogenesis involved in innervation
          alveolar secondary septum development
          lens fiber cell development
          positive regulation of ERK1 and ERK2 cascade
          bone maturation
          epithelial cell fate commitment
          positive regulation of MAPKKK cascade by fibroblast growth factor receptor signaling pathway
          cochlea development
          positive regulation of canonical Wnt signaling pathway
          fibroblast growth factor receptor apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          fibroblast growth factor-activated receptor activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          fibroblast growth factor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0