Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GNB2L1 SIRNA Assays ›
          Gene Symbol:
          Gnb2l1
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 2 like 1
          Gene Aliases:
          AL033335, GB-like, Gnb2-rs1, Rack1, p205
          Chromosome Location:
          Chr.11: 48800360 - 48806241 on Build GRCm38
          UniGene ID:
          Mm.5305
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62042

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Gnb2l1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008143.3 NP_032169.1 7 898
          GenBank mRNA AK002493.1 - 7 887
          AK012242.1 - 6 685
          AK017772.1 - 7 899
          AK051226.1 - 7 899
          AK159797.1 - 7 899
          AK160528.1 - 7 869
          AK160739.1 - 7 896
          AK161234.1 - 7 899
          AK166417.1 - 7 887
          AK166786.1 - 7 901
          AK166800.1 - 7 902
          AK166889.1 - 6 539
          AK166945.1 - 7 899
          AK168086.1 - 7 899
          AK168096.1 - 7 899
          AK168196.1 - 7 899
          AK168349.1 - 7 930
          AK169612.1 - 7 898
          AK186349.1 - 2 62
          BC020322.1 - 7 875
          BC036295.1 - 5 438
          BC046760.1 - 7 883 5693613
          D29802.1 - 7 892
          X75313.1 - 7 898

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_130734, FQ213067, FQ220598, FQ229908, FQ218475, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          positive regulation of protein phosphorylation
          regulation of translation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cell cycle
          multicellular organism development
          gastrulation
          protein localization
          negative regulation of gene expression
          negative regulation of translation
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of cAMP catabolic process
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of protein homooligomerization
          negative regulation of peptidyl-serine phosphorylation
          intracellular signal transduction
          regulation of growth
          positive regulation of apoptotic process
          positive regulation of GTPase activity
          rhythmic process
          negative regulation of phagocytosis
          positive regulation of cyclic-nucleotide phosphodiesterase activity
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          positive regulation of mitochondrial depolarization
          negative regulation of protein tyrosine kinase activity
          cellular response to glucose stimulus
          regulation of establishment of protein localization to plasma membrane
          negative regulation of hydrogen peroxide-induced neuron death
          positive regulation of intrinsic apoptotic signaling pathway
          receptor activity
          protein kinase C binding
          protein binding
          ion channel inhibitor activity
          cysteine-type endopeptidase activator activity involved in apoptotic process
          enzyme binding
          protein phosphatase binding
          protein tyrosine kinase inhibitor activity
          receptor tyrosine kinase binding
          signaling adaptor activity
          SH2 domain binding
          protein homodimerization activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0