Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FOXA1 SIRNA Assays ›
Gene Symbol:
Foxa1
Gene Name:
forkhead box A1
Gene Aliases:
Hnf-3a, Hnf3a, Tcf-3a, Tcf3a
Chromosome Location:
Chr.12: 57540628 - 57546911 on Build GRCm38
UniGene ID:
Mm.4578
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
62135
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Foxa1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008259.3
NP_032285.2
2
1766
GenBank mRNA
BC096524.1
-
2
1766
5720113
U44752.1
-
2
1607
X74936.1
-
2
1573
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_004496, AK316360, BC033890, U39840, AK313785.
This siRNA also targets the following transcripts in Rat: NM_012742, X55955.
Panther Classification:
Molecular Function -
winged helix/forkhead transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
Notch signaling pathway
multicellular organism development
regulation of gene expression
negative regulation of epithelial to mesenchymal transition
chromatin modification
dorsal/ventral neural tube patterning
lung development
response to estradiol
positive regulation of intracellular estrogen receptor signaling pathway
tube morphogenesis
hormone metabolic process
glucose homeostasis
positive regulation of neuron differentiation
positive regulation of smoothened signaling pathway
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
anatomical structure formation involved in morphogenesis
neuron fate specification
positive regulation of sequence-specific DNA binding transcription factor activity
regulation of cell cycle
epithelial tube branching involved in lung morphogenesis
lung epithelial cell differentiation
secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
epithelial-mesenchymal signaling involved in prostate gland development
prostate gland epithelium morphogenesis
prostate gland stromal morphogenesis
epithelial cell maturation involved in prostate gland development
connective tissue development
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor binding
protein domain specific binding
sequence-specific DNA binding
transcription regulatory region DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0