Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ITGAV SIRNA Assays ›
          Gene Symbol:
          Itgav
          Gene Name:
          integrin alpha V
          Gene Aliases:
          1110004F14Rik, 2610028E01Rik, CD51, D430040G12Rik
          Chromosome Location:
          Chr.2: 83723819 - 83806917 on Build GRCm38
          UniGene ID:
          Mm.227
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62430

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Itgav
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008402.3 NP_032428.2 1 481
          GenBank mRNA AK020674.1 - 1 413
          AK080357.1 - 1 420
          AK140647.1 - 1 422
          AK149984.1 - 1 383
          BB632484.1 - 1 482
          BC021600.1 - 1 351
          BC133682.1 - 1 193
          U14135.1 - 1 363

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: XM_008762000.

          Panther Classification:

          Molecular Function -

          integrin

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          blood vessel development
          cellular calcium ion homeostasis
          inflammatory response
          cell adhesion
          cell-matrix adhesion
          positive regulation of cytosolic calcium ion concentration
          integrin-mediated signaling pathway
          multicellular organism development
          positive regulation of cell proliferation
          fertilization
          negative regulation of macrophage derived foam cell differentiation
          negative regulation of lipid storage
          cell growth
          cell migration
          cell differentiation
          positive regulation of cell migration
          cell-substrate adhesion
          negative regulation of lipid transport
          detection of molecule of bacterial origin
          positive regulation of osteoblast proliferation
          heterotypic cell-cell adhesion
          substrate adhesion-dependent cell spreading
          regulation of ion transmembrane transport
          gonad morphogenesis
          endodermal cell differentiation
          apolipoprotein A-I-mediated signaling pathway
          T cell activation
          apoptotic cell clearance
          positive regulation of MAPK cascade
          endothelial cell migration
          regulation of bone resorption
          negative regulation of low-density lipoprotein particle receptor biosynthetic process
          positive regulation of cell adhesion
          viral entry into host cell
          focal adhesion assembly
          negative regulation of lipoprotein metabolic process
          regulation of phagocytosis
          negative chemotaxis
          release of sequestered calcium ion into cytosol
          trophoblast giant cell differentiation
          ERK1 and ERK2 cascade
          calcium ion transmembrane transport
          response to nitric oxide
          extrinsic apoptotic signaling pathway in absence of ligand
          regulation of apoptotic cell clearance
          negative regulation of entry of bacterium into host cell
          negative regulation of extrinsic apoptotic signaling pathway
          opsonin binding
          fibronectin binding
          protease binding
          protein kinase C binding
          receptor binding
          voltage-gated calcium channel activity
          insulin-like growth factor I binding
          peptide binding
          metal ion binding
          protein heterodimerization activity
          extracellular matrix binding
          extracellular matrix protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0