Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KCNQ1 SIRNA Assays ›
          Gene Symbol:
          Kcnq1
          Gene Name:
          potassium voltage-gated channel, subfamily Q, member 1
          Gene Aliases:
          AW559127, KVLQT1, Kcna9
          Chromosome Location:
          Chr.7: 143106742 - 143427050 on Build GRCm38
          UniGene ID:
          Mm.439769
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62555

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Kcnq1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008434.2 NP_032460.2 7 1064
          GenBank mRNA AB079603.1 - 7 767
          AK154244.1 - 7 1038
          AY331142.1 - 7 979
          AY644390.1 - 2 172
          BC045142.1 - 7 1064 6314642
          BC055304.1 - 7 1019 6397978
          U70068.1 - 7 870

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of defense response to virus by host
          regulation of gene expression by genetic imprinting
          transport
          ion transport
          potassium ion transport
          positive regulation of heart rate
          gene silencing
          regulation of ion transmembrane transport
          cellular response to drug
          regulation of membrane potential
          negative regulation of insulin secretion
          transmembrane transport
          cardiac muscle contraction
          regulation of membrane repolarization
          regulation of ventricular cardiac muscle cell membrane repolarization
          regulation of atrial cardiac muscle cell membrane repolarization
          positive regulation of cardiac muscle contraction
          positive regulation of gastric acid secretion
          cellular response to cAMP
          potassium ion export
          potassium ion transmembrane transport
          response to anesthetic
          cardiovascular system development
          ventricular cardiac muscle cell action potential
          membrane repolarization during action potential
          membrane repolarization during cardiac muscle cell action potential
          atrial cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          potassium ion export across plasma membrane
          positive regulation of potassium ion transmembrane transport
          ion channel activity
          voltage-gated ion channel activity
          voltage-gated potassium channel activity
          delayed rectifier potassium channel activity
          potassium channel activity
          calmodulin binding
          protein phosphatase 1 binding
          outward rectifier potassium channel activity
          protein kinase A catalytic subunit binding
          protein kinase A regulatory subunit binding
          protein homodimerization activity
          ion channel binding
          voltage-gated potassium channel activity involved in cardiac muscle cell action potential repolarization
          voltage-gated potassium channel activity involved in atrial cardiac muscle cell action potential repolarization
          scaffold protein binding
          voltage-gated potassium channel activity involved in ventricular cardiac muscle cell action potential repolarization

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0