Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110315.2 | NP_001103785.1 | 4 | 368 | ||
NM_001294149.1 | NP_001281078.1 | 4 | 368 | |||
NM_001294150.1 | NP_001281079.1 | 3 | 256 | |||
NM_008440.4 | NP_032466.2 | 4 | 368 | |||
GenBank mRNA | AK138237.1 | - | 4 | 310 | ||
AK147640.1 | - | 4 | 326 | |||
AY426748.1 | - | 3 | 197 | |||
BC034863.1 | - | 4 | 313 | |||
BC045161.1 | - | 4 | 341 | |||
BC054538.1 | - | 4 | 341 | |||
BC062891.1 | - | 4 | 347 | 5702372 | ||
CJ103366.1 | - | 4 | 369 | |||
CJ115414.1 | - | 4 | 360 | |||
D29951.1 | - | 4 | 317 |